Lipofectamine™ LTX Reagent with PLUS™ Reagent
See the new Lipofectamine® LTX & Plus Reagent protocol—now optimized for efficiency, viability and reproducibility
Lipofectamine™ LTX Reagent with PLUS™ Reagent
Invitrogen™

Lipofectamine™ LTX Reagent with PLUS™ Reagent

Lipofectamine™ LTX Reagent is a plasmid transfection reagent that offers the right balance of potency and gentleness for your cells,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
153380300.3 mL
A126210.1 mL
153381001 mL
Catalog number 15338030
Price (USD)/ Each
270.00
Special Offer
Online exclusive
Ends: 15-Mar-2025
300.00 
Save 30.00 (10%)
-
Add to cart
Quantity:
0.3 mL
Price (USD)/ Each
270.00
Special Offer
Online exclusive
Ends: 15-Mar-2025
300.00 
Save 30.00 (10%)
Add to cart
Lipofectamine™ LTX Reagent with PLUS™ Reagent
Catalog number15338030
Price (USD)/ Each
270.00
Special Offer
Online exclusive
Ends: 15-Mar-2025
300.00 
Save 30.00 (10%)
-
Add to cart
Lipofectamine™ LTX Reagent is a plasmid transfection reagent that offers the right balance of potency and gentleness for your cells, resulting in the highest possible transfection efficiencies and viabilities. Lipofectamine™ LTX Reagent is an advanced solution for gene expression in both common and challenging cell types. No other plasmid DNA-specific transfection reagent can match the efficiency, convenience, and gentleness of Lipofectamine™ LTX Reagent for transfection of primary, challenging, and sensitive cell lines. Lipofectamine™ LTX Reagent is synthesized from 100% animal origin-free components, making it easy to validate the absence of zoonotic diseases, such as BSE or viruses, in research experiments or cell lines.

Lipofectamine™ LTX Reagent offers:

• Superior delivery—transfect a broad range of cell types, including primary, challenging, and sensitive cells, with high efficiency
• Superior performance—combine high transfection efficiency with maximal (>90%) cell viability
• Optimized protocols—rapid, simple protocols provided for most cell lines
• A highly effective reagent to generate stable cell lines

Transfect primary cells efficiently
Primary cells are often difficult to transfect. Lipofectamine™ LTX Reagent, when combined with PLUS™ Reagent, provides the highest level of transfection in primary cells of any lipid- or polymer-based reagent. Many primary cell types have been successfully transfected with Lipofectamine™ LTX Reagent, including primary neural progenitor cells.

Superior delivery into challenging cells
When it comes to achieving high levels of protein expression in RAW267.3 macrophages, Lipofectamine™ LTX Reagent outperforms competing transfection reagents (see figure). Achieving high-level transgene expression in cells that are traditionally refractory to transfection allows you to obtain results that were previously unattainable.

Broad compatibility with cell lines
In addition to being excellent for transfecting primary and challenging cells, Lipofectamine™ LTX Reagent provides you with the most versatile approach to transfecting established cells lines, including those that are sensitive to other transfection reagents. Use our Transfection Selection Tool to help you select the best transfection conditions for the cells with which you work.

Stable transfection
Lipofectamine™ LTX strikes the right balance between potency and gentleness, giving you the highest possible percentage of successfully transfected cells. This makes Lipofectamine™ LTX Reagent the quickest and easiest reagent for establishing stable transfectants from challenging or refractory cell types.

Simple, rapid, serum-compatible protocol
Lipofectamine™ LTX Reagent offers a streamlined protocol—no need to remove transfection complexes or change/add medium following transfection. A simple optimization protocol helps you get the best results quickly, so you'll spend less hands-on time, and get faster results.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free
Cell TypeEstablished Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample TypePlasmid DNA, RNAi Plasmids (shRNA, miR)
Transfection TechniqueLipid-based Transfection
For Use With (Application)Transfection
High-throughput CompatibilityHigh-throughput Compatible
Product LineLipofectamine™
Product TypeTransfection Reagent
Quantity0.3 mL
Serum CompatibleYes
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
Each unit contains one vial of Lipofectamine™ LTX Reagent (0.3 ml) and one vial of Plus Reagent (250 μl). Store at 2–8°C.