Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well |
Detection Method | Colorimetric |
Packaging | 5 Plates |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 200 μL |
No. of Wells | 96 |
Surface Treatment | Divalent Nickel, Metal Chelate |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Black, 96-well |
Detection Method | Fluorescence |
Packaging | 5 Plates |
Product Line | Pierce |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 200 μL |
No. of Wells | 96 |
Surface Treatment | Divalent Nickel, Metal Chelate |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip |
Detection Method | Colorimetric |
Packaging | 5 Plates |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 200 μL |
No. of Wells | 96 |
Surface Treatment | Divalent Nickel, Metal Chelate |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | White, 96-well |
Detection Method | Chemiluminescence |
Packaging | 5 Plates |
Product Line | Pierce |
Color | White |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 200 μL |
No. of Wells | 96 |
Surface Treatment | Divalent Nickel, Metal Chelate |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Catalog Number | Specifications | Unit Size | Description | Detection Method | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
15442 | 5 plates | Clear, 96-well | Colorimetric | 269.65 Online price 283.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15342 | 5 plates | Black, 96-well | Fluorescence | 285.65 Online price 300.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15142 | 5 plates | Clear, 8-well Strip | Colorimetric | 277.65 Online price 293.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15242 | 5 plates | White, 96-well | Chemiluminescence | 285.65 Online price 300.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|