Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:For PCR enzyme with ≤0.01 copy of bacterial gDNA per enzyme unit, check out Invitrogen Platinum Taq DNA Polymerase, DNA-free.
Invitrogen™

Taq DNA Polymerase, native

Catalog number: 18038042
Have Questions?
Invitrogen™

Taq DNA Polymerase, native

Catalog number: 18038042
Have Questions?
Catalog Number
18038042
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3 x 500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
18038042500 units
-
18038018100 units
-
180380673 x 500 units
-
180382405,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. The enzyme consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5´→3´ DNA polymerase activity and a 5´→3´ exonuclease activity (see figure). With Taq DNA Polymerase, you get:

  • Your choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • An enzyme that is licensed and qualified for PCR

    Applications
    Amplification of DNA from complex genomic, viral, and plasmid templates, RT-PCR, sequencing ssDNA, and cycle sequencing

    Source
    Native enzyme is purified from Thermus aquaticus YT1.

    Unit definition
    One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 min at 74°C.
  • For Research Use Only. Not for use in diagnostic procedures.

    Specifications

    Detection Method
    Primer-probe
    Format
    Stand-alone enzyme
    GC-Rich PCR Performance
    Low
    PCR Method
    qPCR, Standard PCR
    Polymerase
    Taq DNA Polymerase
    Reaction Speed
    Standard
    Exonuclease Activity
    5' - 3'
    Product Type
    DNA Polymerase
    Quantity
    500 Units
    Shipping Condition
    Approved for shipment on Wet or Dry Ice
    Concentration
    5 U/μL
    Fidelity (vs. Taq)
    1 X
    Hot Start
    No
    No. of Reactions
    500
    Overhang
    3'-A
    Reaction Format
    Separate Components
    Size (Final Product)
    5 kb or less
    Starting Material
    DNA

    Contents & Storage

    Contains:
    • 100 μL Taq DNA Polymerase (5 U/μL)
    • 2.5 mL 10X PCR buffer (without magnesium)
    • 1 ml Magnesium Chloride (50 mM)

    Store at -20°C in a non-frost-free freezer. Stable for 6 months when stored properly.

    Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text