Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:For superior cDNA synthesis performance upgrade to Invitrogen™ SuperScript™ IV Reverse Transcriptase.
Invitrogen™

SuperScript™ II Reverse Transcriptase

Catalog number: 18064014
Have Questions?
Invitrogen™

SuperScript™ II Reverse Transcriptase

Catalog number: 18064014
Have Questions?
Catalog Number
18064014
Unit Size
10,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 10,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1806401410,000 units
-
180640222,000 units
-
180640714 x 10,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT. Mutations in the RNase H domain of the enzyme eliminate degradation of the RNA during first-strand cDNA synthesis, which results in higher yields of full-length cDNA.

SuperScript reverse transcriptases are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.

Features of SuperScript II Reverse Transcriptase include:
• High cDNA yields and more full-length product
• Generation of cDNA up to 12.3 kb
• First-strand cDNA synthesis at higher temperatures than conventional MMLV RT

Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase, features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Stand-alone Enzyme
GC-Rich PCR Performance
High
Reaction Speed
50 min.
Technique
Reverse Transcription
Optimal Reaction Temperature
42°C
Quantity
10,000 Units
Reverse Transcriptase
SuperScript II
Ribonuclease H Activity
Reduced
Shipping Condition
Wet Ice
Concentration
200 U/μL
Final Product Type
First-Strand cDNA
No. of Reactions
50 Reactions
Reaction Format
Separate components
Reagent Type
Reverse Transcription
Size (Final Product)
Up to 12.3 kb
Starting Material
RNA

Contents & Storage

• SuperScript II Reverse Transcriptase, 50 μL (10,000 units at 200 U/μL)
• 5X First-strand buffer
• 100 mM DTT

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text