M13KO7 Helper Phage is suitable for use with M13/phagemid hosts for the production of ssDNA from vectors containing an f1orRead more
Have Questions?
Catalog Number
Quantity
18311019
5 x 1 mL
Catalog number 18311019
Price (USD)
333.65
Online Exclusive
355.00
Save 21.35 (6%)
Each
Add to cart
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)
333.65
Online Exclusive
355.00
Save 21.35 (6%)
Each
Add to cart
M13KO7 Helper Phage is suitable for use with M13/phagemid hosts for the production of ssDNA from vectors containing an f1or f1-like origin of replication. Prepared by a modification of the procedure of Vieira and Messing (1), this high-titer helper phage eliminates the need to make and maintain a separate stock of helper phage. It contains the Kmr gene and a p15A origin of replication that allows it to replicate like a plasmid inside the cells. Detailed protocols for the production of ss DNA are provided on the product profile. M13KO7 Helper Phage offers:
• Convenient preparation of ss DNA from M13 or phagemid vectors • >1 x 1011 pfu/ml
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeM13KO7 Helper Phage
Quantity5 x 1 mL
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store in ultra-cold freezer (-68 to -85°C).
Frequently asked questions (FAQs)
Do you offer an alternative for M13KO7 Helper Phage (Cat. No. 18311019)?
We do not offer an alternative for M13KO7 Helper Phage.