Informational:Culturing neurons? Try our new Gibco™ Poly-D-Lysine Solution.
Informational:Increase cell survival by more than 50% with new Gibco™ B-27™ Plus Neuronal Culture System »
Gibco™

Neurobasal™ Medium

Catalog number: 21103049
Have Questions?
Gibco™

Neurobasal™ Medium

Catalog number: 21103049
Have Questions?
Catalog Number
21103049
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
21103049500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Neurobasal Medium is a basal medium designed for long-term maintenance and maturation of pure pre-natal and embryonic neuronal cell populations without the need for an astrocyte feeder layer when used with Gibco B-27 supplements.

Neurobasal Medium is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of Neurobasal media and B-27 supplements.

For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Neural Cells
Classification
Serum-free
Culture Type
Neuronal Cell Culture
Form
Liquid
Product Type
Neurobasal Medium
Shelf Life
12 Months From Date of Manufacture
With Additives
Phenol Red
Product Line
Neurobasal™
Shipping Condition
Ambient
Tested For
Endotoxin, Performance, Sterility, Osmolality, pH

Contents & Storage

Store in refrigerator (2–8°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text