1x1 image pixel for data collection
Pierce™ Concentrated Buffer Stocks (10X and 20X)
Pierce™ Concentrated Buffer Stocks (10X and 20X)
Thermo Scientific™

Pierce™ Concentrated Buffer Stocks (10X and 20X)

Save time using Pierce Concentrated Buffer stock solutions (10X and 20X) with simple dilutions to expedite your experiment without sacrificing quality or consistency.
Promo ImagePromotion
Promo code:P5934129
Save across your therapeutic antibody development workflow
Get up to 34% off products for your therapeutic antibody development workflow, including cloning, purification, antibody labeling, and more! Take advantage of our comprehensive solutions and maximize your efficiency. Offer ends August 31, 2025.Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentrationProduct Type
28360Promo Image500 mL20XTBS Tween 20 Buffer
28352Promo Image500 mL20XPBS Tween 20 Buffer
28358Promo Image500 mL20XTBS Buffer
28348Promo Image500 mL20XPhosphate Buffered Saline (PBS)
28362Promo Image1 L10XTris-Glycine SDS Buffer
28341500 mL20XBorate Buffer
28344500 mL20XDPBS
28363Promo Image1 L10XTris-Glycine Buffer
28355Promo Image1 L10XTBE Buffer
28346500 mL20XDPBS Tween 20 Buffer
28354Promo Image500 mL20XTAE Buffer
Catalog number 28360
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
500 mL
Concentration:
20X
Product Type:
TBS Tween 20 Buffer
Request bulk or custom format
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Add to cart
Our variety of Pierce Concentrated Buffer stock solutions are ready to use without having to weigh and dissolve dry ingredients or adjust the pH with concentrated acid or base. Simply dilute the liquid stock solution with pure water and proceed with your experiment.

Benefits of using Pierce Concentrated Buffer stock solutions include:
Convenient—no weighing, no pH adjustment, no need to stock individual components
Improved accuracy—ready-to-use formula, easily diluted without weighing chemical components
Consistent quality—our quality control ensures that every prep will yield the same, consistent buffer

20X PBS Tween-20 can be used to prepare PBS-Tween (PBS-T) wash buffers for ELISA, western, and other immunoassays, as well as blocking buffer for plate-based assays. When diluted to 1X with water, 20X PBS Tween-20 makes 10 mM sodium phosphate, 0.15 M NaCl, 0.05% Tween™ 20 buffer at pH 7.5.

20X TBS Tween-20 (Tris-buffered saline with Tween™ 20 detergent) can be used to prepare TBS-Tween (TBS-T) wash buffers or blocking buffer diluent for applications such as ELISA, western blotting, and other immunoassays. When diluted 20-fold in water, a TBS Tween-20 buffer containing 25 mM Tris, 0.15 M NaCl, 0.05% Tween-20 at pH 7.5 is ready for use.

20X Tris-Buffered Saline (TBS) is a stock solution for preparing Tris-NaCl buffer for use as a wash buffer and antibody diluent for ELISA, western blotting, and other immunoassays. 20X Tris Buffered Saline (TBS) makes 25 mM Tris, 0.15 M NaCl with pH 7.2 to 7.5 when diluted to 1X with water.

20X Phosphate-Buffered Saline (PBS) is ideal for preparing PBS buffers for use in crosslinking, biotinylation, and fluorescent labeling reactions requiring an amine-free buffer. When diluted 20-fold in water, the solution yields 10 mM sodium phosphate, 0.15 M NaCl at pH 7.5.

10X Tris-Glycine-SDS Buffer makes 0.025 M Tris, 0.192M glycine, 0.1% sodium dodecyl sulfate (SDS) at pH 8.5 when diluted to 1X with water.

20X Borate Buffer is ideal for preparing sodium borate buffer solutions for use in protein modification procedures requiring amine-free buffer at an alkaline pH. 20X Borate Buffer makes 50 mM borate at pH 8.8 when diluted to 1X with water.

20X Modified Dulbecco's PBS is used for preparing physiological Na- and K-phosphate buffered saline (D-PBS) used for wash buffers and antibody diluents in immunoassays like ELISA and western blot. 20X Modified Dulbecco's PBS makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl, pH 7.4 when diluted to 1X with water.

10X Tris-Glycine Buffer is a space-saving stock solution that is ideal for quickly preparing standard Tris-glycine (pH 8.5) transfer buffer used for western blotting using Tris-glycine gel electrophoresis. When diluted 10-fold in water or 20% methanol, the solution yields 0.025 M Tris, 0.192 M glycine at pH 8.5.

10X TBE Buffer stock solution is ideal for casting and preparing Tris-borate-EDTA (TBE) running buffer used for agarose and polyacrylamide gel electrophoresis of nucleic acids. When diluted to 1X, the TBE Buffer contains 0.089 M Tris, 0.089 M borate, 2 mM EDTA with pH 8.2 to 8.4.

20X Modified Dulbecco's PBS Tween-20 will make physiological sodium- and potassium-phosphate buffered saline (D-PBS) to be used as a wash buffer for ELISA, western, and other immunoassays or as diluent or blocking buffer for plate-based assays. 20X Modified Dulbecco's PBS Tween-20 (D-PBS-T) makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl, 0.05% Tween™ 20 at pH 7.4 when diluted to 1X with water.

20X TAE Buffer is a space-saving stock solution that is ideal for casting and preparing Tris-acetate EDTA (TAE) running buffer for agarose gel electrophoresis of nucleic acids. When diluted to 1X with water, a 0.04 M Tris, 0.04 M acetate, 1 mM EDTA buffer at pH 8.2 to 8.4 is ready for use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20X
Product LinePierce™, Tween™
Quantity500 mL
FormLiquid
Product TypeTBS Tween 20 Buffer
pH7.5
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20–25°C.

Frequently asked questions (FAQs)

What is the optimal pH for the Pierce Concentrated Buffer Stocks (10X and 20X) (Cat. No. 28362)?

The quality specifications for the ready to use 1x solution is 8.5 ± 0.2. (see example COA: https://assets.thermofisher.com/TFS-Assets/LSG/certificate/Certificates-of-Analysis/28362_WH332690.PDF). The pH should be between 8.3 and 8.7.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can you provide the shelf-life for the Pierce 20X TBS Tween 20 Buffer?

The Pierce 20X TBS Tween 20 Buffer is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link