Thermo Scientific™
OPD (o-phenylenediamine dihydrochloride)
Catalog number: 34005
Have Questions?
Thermo Scientific™
OPD (o-phenylenediamine dihydrochloride)
Catalog number: 34005
Have Questions?
Catalog Number
34005
Unit Size
25 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
3400525 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce OPD is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
OPD yields a water soluble yellow-orange product when reacted with peroxidase with an absorbance maximum of 492. OPD can easily be dissolved in a substrate buffer such as Stable Peroxide Substrate Buffer (Part No. 34062).
Related Products
OPD Substrate Tablets (o-phenylenediamine dihydrochloride)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Pierce OPD Powder
Product Type
ELISA Substrate
Detection Method
Colorimetric
Substrate Properties
Chemical Substrate
Substrate Type
HRP (Horseradish Peroxidase)
Contents & Storage
Upon receipt store product at -20°C. Product is shipped at ambient temperature.