SuperBlock™ Blocking Buffer
Thermo Scientific™

SuperBlock™ Blocking Buffer

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityChemical Name or Material
375361 LSuperBlock T20 (TBS)
37580100 mLSuperBlock (PBS)
375151 LSuperBlock (PBS)
375185 LSuperBlock (PBS)
37581100 mLSuperBlock (TBS)
375351 LSuperBlock (TBS)
375455 Pk.SuperBlock (TBS) Dry Blend
375161 LSuperBlock T20 (PBS)
Catalog number 37536
Price (USD)
251.65
Online Exclusive
303.00
Save 51.35 (17%)
Each
Quantity:
1 L
Chemical Name or Material:
SuperBlock T20 (TBS)
Request bulk or custom format
Price (USD)
251.65
Online Exclusive
303.00
Save 51.35 (17%)
Each

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.

Features of SuperBlock Blocking Buffer include:

  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialSuperBlock T20 (TBS)
Recommended StorageUpon receipt store at 4°C. Product is shipped at ambient temperature.
Concentration1X
For Use With (Application)ELISA
Physical FormLiquid
Product LineSuperBlock
Quantity1 L
Unit SizeEach

Frequently asked questions (FAQs)

What should I do about the colloidal carbon (black precipitate) in SuperBlock Blocking Buffer solution?

Allow any colloidal carbon, if present, to settle to the bottom of the bottle and pouring off what you need to use. We do not recommend centrifugation or filtering because these procedures may reduce the product's blocking capabilities.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What preservative is used in SuperBlock Blocking Buffer, and what is its source?

Kathon Preservative, a broad spectrum antimicrobial agent, is used at a concentration of 600 ppm.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is SuperBlock Blocking Buffer's shelf life?

Its shelf life is one year at 4°C.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What incubation time and temperature should I use for effective blocking using the SuperBlock Blocking Buffer ?

Typical procedures use 1 hour at room temperature or overnight at 4°C. However, sufficient blocking in many procedures is possible in 3 x 2 minutes at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Is SuperBlock Blocking Buffer RNase-free?

No, SuperBlock Blocking Buffer is not RNase-free.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.