Thermo Scientific™

1-Step™ ABTS Substrate Solution

Catalog number: 37615
Have Questions?
Thermo Scientific™

1-Step™ ABTS Substrate Solution

Catalog number: 37615
Have Questions?
Catalog Number
37615
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
37615250 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
ABTS Substrate is a water-soluble peroxidase substrate that yields a measurable green end product for use in ELISA methods.

ABTS (2,2'-Azinobis [3-ethylbenzothiazoline-6-sulfonic acid]-diammonium salt) is a water-soluble HRP substrate that yields a green end product upon reaction with peroxidase. The green product has two major absorbance peaks, 410nm and 650nm. ABTS is less sensitive than OPD and TMB in ELISA applications. It is less readily oxidized, and its color development is slower (approximately 20 minutes). This may be advantageous if unacceptable background results from the use of the OPD or TMB substrates due to higher sensitivities.

Related Products
ABTS Substrate Tablets
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce 1-Step ABTS Solution
Product Type
ELISA Substrate
Detection Method
Colorimetric
Product Line
1-Step™
Substrate Properties
Chemical Substrate
Substrate Type
HRP (Horseradish Peroxidase)

Contents & Storage

Store at 4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text