Specifications
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
PCR Method2-step RT-qPCR
Reaction SpeedFast
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Optimal Reaction Temperature42°C
Product LineTaqMan™
Product TypeMicroRNA Reverse Transcription Kit
Quantity200 reactions
Reverse TranscriptaseMultiScribe™
Shipping ConditionDry Ice
Sufficient For200 Reactions
For Use With (Application)miRNA analysis
Concentration10X
Fidelity (vs. Taq)10 X
Final Product TypecDNA
No. of Reactions200 Reactions
Reaction FormatSeparate Components
Sample TypeRNA
Size (Final Product)200 bp or less
Unit SizeEach