Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

TaqMan™ Cells-to-CT™ Control Kit

Catalog number: 4386995
Have Questions?
Invitrogen™

TaqMan™ Cells-to-CT™ Control Kit

Catalog number: 4386995
Have Questions?
Catalog Number
4386995
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4386995100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ambion™ TaqMan™ Cells-to-CT Control Kit is designed for use with the TaqMan™ Gene Expression Cells-to-CT Kit. The Control Kit includes XenoRNA™ Control, a synthetic RNA transcript with a unique sequence that lacks homology to current annotated biological sequences, making it an ideal control for any experiment. The Control Kit also includes a TaqMan™ Gene Expression Assay for the XenoRNA™ Control target, and a TaqMan™ Gene Expression Assay for the highly expressed endogenous control gene β-actin.

Features of the The Ambion™ TaqMan™ Cells-to-CT Control Kit:
• Confirms sufficient cell input
• Monitors the presence of RT or real-time PCR inhibitors
• Controls for sample normalization

Monitor Inhibition and Confirm Sufficient Cell Input
The TaqMan™ Cells-to-CT™ Control Kit is used to assess inhibition and cell input in Cells-to-CT reactions. For example, the XenoRNA™ Control confirms that samples can support RT-PCR and is an indicator of reaction inhibitors. By adding the XenoRNA™ Control to the Stop Solution during the last step of the cell lysis procedure, each sample is provided with a constant amount of control target and subsequently amplified by the XenoRNA™ TaqMan™ Gene Expression Assay.

Amplification with the β-actin Gene Expression Assay functions as an endogenous control for normalization, and ensures that the lysis reaction contains a sufficient number of cells—this is important for experiments with fewer than ∼100 cells and monitors the effectiveness of cell lysis.

Evaluate Compatibility with New Cell Lines
The TaqMan™ Cells-to-CT™ Control Kit is ideal for assessing the compatibility of untested cell lines with the TaqMan™ Gene Expression Cells-to-CT Kit to determine the maximum number of cells that can be used per lysis reaction.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Template
XenoRNA™ control (synthetic RNA transcript)
Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Gene Symbol
ACTB
Green Features
Fewer resources used and less waste, Less hazardous
Internal Probe Modification
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye
FAM
Product Line
Ambion™, Cells-to-CT™, TaqMan™
Product Type
Cells-to-C Control Kit
Shipping Condition
Dry Ice
Sufficient For
100 Reactions
Target
XenoRNA™ control, β-actin
For Use With (Application)
Gene Expression
No. of Reactions
100 Reactions

Contents & Storage

Xeno™ RNA Control, 20X Xeno™ RNA Control Taqman™ Gene Expression Assay, and 20X ACTB (β-actin) TaqMan™ Gene Expression Assay are all stored at -20°C.

All reagents are guaranteed stable for 6 months when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text