Pierce™ F(ab')2 Micro Preparation Kit
Pierce™ F(ab')2 Micro Preparation Kit
Thermo Scientific™

Pierce™ F(ab')2 Micro Preparation Kit

The Thermo Scientific Pierce F(ab')2 Micro Preparation Kit uses immobilized pepsin protease to selectively digest whole human and other IgGRead more
Have Questions?
Catalog number 44688
Price (USD)/ Each
540.65
Online exclusive
578.00 
Save 37.35 (6%)
-
Add to cart
Quantity:
10 Antibody Samples, each containing 25 to 250 μg IgG
Request bulk or custom format
Price (USD)/ Each
540.65
Online exclusive
578.00 
Save 37.35 (6%)
Add to cart
Pierce™ F(ab')2 Micro Preparation Kit
Catalog number44688
Price (USD)/ Each
540.65
Online exclusive
578.00 
Save 37.35 (6%)
-
Add to cart
The Thermo Scientific Pierce F(ab')2 Micro Preparation Kit uses immobilized pepsin protease to selectively digest whole human and other IgG antibodies to make F(ab')2 fragments that retain antigen binding activity.

Features of the F(ab')2 Micro Preparation Kit:

Ideal when sample size is limited—Use the micro kit for 25 to 250 μg IgG.
Enzyme-free digestion products—Immobilized Pepsin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the proteolytic enzyme
Suitable for human and other species of IgG—the kit procedure is optimized for human, rabbit and mouse IgG. Pepsin-based digestion is effective for other species and subclasses of IgG, although the purification step requires that the antibody effectively binds to Protein A. (Note: for best results with mouse IgG1, use Part No. 44980.)
Complete—kits include all reagents needed to prepare and purify antibody fragments
Fast—spin format greatly reduces sample processing time
Flexible—protocols are included for multiple species and IgG subclasses, as well as sample size and concentration.
Efficient—enhanced yield and sample purity

These F(ab')2 Preparation Kits are suitable for human, rabbit, mouse and other species and subclasses of IgG except mouse IgG1. The antibody digestion reaction with pepsin agarose is performed in convenient disposable spin columns that allow efficient removal of the immobilized protease and maximum recovery of the IgG fragments. Also included in the kits are Thermo Scientific NAb Protein A Spin Columns and buffers to efficiently purify the resulting fragments. Protein A binds the Fc fragments and undigested IgG, allowing the pure F(ab')2 fragments to be recovered in the flow-through fraction. The kits also include Thermo Scientific Zeba Desalting Spin Columns for preparing the IgG sample quickly without dilution instead of utilizing time-consuming dialysis steps.

The kits use pepsin, a nonspecific endopeptidase, to enzymatically digest the Fc portion of whole IgG to yield the fragment known as F(ab')2. This fragment is composed of a pair of Fab' units connected by two disulfide bonds. Because the pepsin protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free.

Related Products
Pierce™ F(ab')2 Preparation Kit
Pierce™ Fab Micro Preparation Kit
Pierce™ Mouse IgG1 Fab and F(ab')2 Micro Preparation Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce F(ab')2 Micro Preparation Kit
Quantity10 Antibody Samples, each containing 25 to 250 μg IgG
Product LinePierce™
Target GeneERK5/BMK1 (pan)
Unit SizeEach
Contents & Storage
Sufficient For: 10 antibody samples, each containing 25 to 250 μg IgG
• Pierce Pepsin Agarose, 0.5 mL
• F(ab')2 Digestion Buffer, 55 mL
• NAb Protein A Plus Spin Columns, 0.2 mL, 2 columns
• PBS Packs (each makes 500 mL), 2 packs
• IgG Elution Buffer, 50 mL
• Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4°C