Search Thermo Fisher Scientific
Catalog Number | For Use With (Application) |
---|---|
60304-201 | Protein Denaturing in Biological Samples |
Thermo Scientific™ HyperSep™ Protein Precipitation Plates provide a quick, effective approach for removal of proteins from biological compounds using the protein crash technique. In combination with SPE and SLE, the Protein Precipitation Plate offers a comprehensive range of options for sample preparation of biological based compounds. The 96-well plates are ideal for use in automated, high throughput systems.
Share catalog number, name or link