Restore™ Fluorescent Western Blot Stripping Buffer
Restore™ Fluorescent Western Blot Stripping Buffer
Thermo Scientific™

Restore™ Fluorescent Western Blot Stripping Buffer

The Thermo Scientific Restore Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primaryRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
62300100 mL
6229920 mL
62300X44 x 100 mL
3 Options
Catalog number 62300
Price (USD)/ 100 mL
183.65
Online exclusive
193.00 
Save 9.35 (5%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ 100 mL
183.65
Online exclusive
193.00 
Save 9.35 (5%)
Add to cart
Restore™ Fluorescent Western Blot Stripping Buffer
Catalog number62300
Price (USD)/ 100 mL
183.65
Online exclusive
193.00 
Save 9.35 (5%)
-
Add to cart
The Thermo Scientific Restore Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primary and near-infrared (IR) dye-labeled secondary antibodies from western blots.

Features of Restore Fluorescent Western Blot Stripping Buffer:

Fast—strip blots in only 15 minutes at room temperature
Saves time—no need to run new gels and prepare a new blot
Conserve samples—reprobe the same PVDF membrane for multiple targets
Economical—less expensive than other commercially available stripping buffers
Efficient—effectively strips blots the first time

Restore Fluorescent Western Blot Stripping Buffer enables the reuse of PVDF membranes, simplifying the Western blot optimization process and allowing the same blot to be reprobed with different primary antibodies to detect alternative targets. Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane only (Part No. 22860).

Fluorescence Western blotting is a powerful method for detecting multiple targets at once. Restore Fluorescent Western Blot Stripping Buffer allows re-probing of PVDF membranes, saving time and cost. This is ideal when samples are limited and optimization or analysis with different primary antibodies is required.

Traditional stripping methods may adversely alter or remove the sample proteins from the PVDF membrane during the stripping process or may be effective for removing only low-affinity antibodies. In contrast, Restore Fluorescent Western Blot Stripping Buffer Stripping efficiency exceeds 90% while reprobing efficiency matches or exceeds other supplier's formulations. Also, Restore Fluorescent Western Blot Stripping Buffer is conveniently stored at room temperature and easy to use. Simply dilute buffer 1:5 in water and incubate your membrane for 5 to 20 minutes at room temperature.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeWestern Blot Stripping Buffer
Detection MethodFluorescence
FormLiquid
For Use With (Application)Western Blot
Membrane CompatibilityPVDF
Product LineRestore™
Quantity100 mL
Unit Size100 mL
Contents & Storage
Upon receipt store at room temperature.