Thermo Scientific™

Mitochondria Isolation Kit for Cultured Cells

Catalog number: 89874
Have Questions?
Thermo Scientific™

Mitochondria Isolation Kit for Cultured Cells

Catalog number: 89874
Have Questions?
Catalog Number
89874
Unit Size
115 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
89874115 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Mitochondria Isolation Kit for Cultured Cells provides a versatile, microcentrifuge-tube method for fractionating intact mitochondria from cultured mammalian cell samples in about 40 minutes.

Features of the Mitochondria Isolation Kit for Cultured Cells:

Fast—isolate intact mitochondria from 20 million cells in approximately 40 minutes (post-cell harvest)
Flexible—process several samples simultaneously using the reagent-based method, or obtain highest possible yield with the Dounce homogenization method
Optimizable—guidelines provided for optimizing purity vs. yield with the reagent-based method
Benchtop-easy—isolation performed in a microcentrifuge tube
Compatible—isolated mitochondria may be lysed with detergent or processed for almost any downstream application, including 2D electrophoresis

Isolation of mitochondria is typically a laborious process requiring single-sample processing with Dounce homogenization. The Mitochondria Isolation Kit uses a non-mechanical, reagent-based method that allows multiple (six) cultured cell samples to be processed concurrently in about 40 minutes. Cultured mammalian cell pellets are gently lysed using a proprietary formulation that results in maximum yield of mitochondria with minimal damage to integrity.

The product instructions describe guidelines for optimizing purity vs. yield parameters. The kit also offers an optimized Dounce homogenization procedure, which results in two-fold greater mitochondria recovery compared to the reagent-based method. Both methods use differential centrifugation to separate the mitochondrial and cytosolic fractions with a benchtop microcentrifuge and are completed in approximately 40 minutes (post-cell harvest). Once isolated, the mitochondria can be used in downstream applications such as apoptosis, signal transduction and metabolic studies, as well as to facilitate mitochondrial proteomics efforts.

Related Products
Mitochondria Isolation Kit for Tissue
Lysosome Enrichment Kit for Tissues and Cultured Cells
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Cell Isolation Kit
Form
Liquid
Reagent Type
Organelle Isolation, Subcellular Fractionation
Sufficient For
50 Isolations
Quantity
115 mL

Contents & Storage

Upon receipt store kit at 4°C. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text