Informational:Taking your work to the clinic? Check out CTS Recombinant Vitronectin »
Gibco™

Vitronectin (VTN-N) Recombinant Human Protein, Truncated

Catalog number: A14700
Have Questions?
Gibco™

Vitronectin (VTN-N) Recombinant Human Protein, Truncated

Catalog number: A14700
Have Questions?
Catalog Number
A14700
Unit Size
1 mL = 0.5 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL = 5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A147001 mL = 0.5 mg
-
A3180410 mL = 5 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8™ Medium (A1517001), vitronectin (VTN-N) has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines. In addition, vitronectin (VTN-N) has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages. As published by Guokai Chen et al. (1) in the laboratory of James Thomson, the VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8™ Medium.

Vitronectin (VTN-N) is:

Optimized—designed in the laboratory of James Thomson for use with Essential 8™ Medium
Defined—for reduced variability in your PSC cultures
Cost effective—for economical and scalable PSC culture

Optimized and Defined Surface
Unlike standard basement membrane extracts (BMEs), vitronectin (VTN-N) was designed in the laboratory of James Thomson for use with Essential 8™ Medium and has been shown to support human pluripotent stem cell attachment and survival better than wild type vitronectin. Vitronectin (VTN-N) provides an optimal environment for feeder-free pluripotent stem cell culture.

Reduce Variability
Vitronectin (VTN-N) is a defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs).

Cost Effective
When used with Essential 8™ Medium, vitronectin (VTN-N) provides a cost effective, defined system for feeder free culture of human pluripotent stem cells (PSCs).

Commercialized in Partnership with Cellular Dynamics International.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
500 μg⁄ml
For Use With (Application)
Stem Cell Culture
Purification Method
HPLC
Quantity
1 mL
Species
Human
Recombinant
Recombinant
Classification
AOF
Endotoxin Level
Very Low
Shipping Condition
Dry ice
Volume (Metric)
1 mL
Protein Form
Recombinant
Form
Liquid
Protein Subtype
Vitronectin
Research Category
Stem Cell Research
Purity or Quality Grade
95 %

Contents & Storage

Store at -80°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text