LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit
LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit
Thermo Scientific™
LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit
The LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential estrogenRead more
Have Questions?
Catalog number A15885
Price (USD)/ 800 x 20 µL assays
2,140.00
-
Add to cart
Price (USD)/ 800 x 20 µL assays
2,140.00
Add to cart
LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit
Catalog numberA15885
Price (USD)/ 800 x 20 µL assays
2,140.00
-
Add to cart
The LanthaScreen™ TR-FRET ER alpha Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential estrogen receptor (ER) alpha ligands as agonists or antagonists of ligand-dependent coactivator recruitment. The kit uses an ER alpha ligand-binding domain (ER alpha-LBD) tagged with glutathione-S-transferase (GST), a terbium (Tb)-labeled anti-GST antibody, and a fluorescein-labeled coactivator peptide in a homogenous mix-and-read assay format.
Agonist Mode
To run the LanthaScreen TR-FRET ER alpha Coactivator Assay in agonist mode (to identify agonist compounds), ER alpha-LBD is added to ligand test compounds, followed by the addition of a mixture of fluorescein-coactivator peptide and Tb-anti-GST antibody. After incubation at room temperature, the TR-FRET 520:495 nm emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound. Based on the biology of the ER alpha-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide.
Antagonist Mode
To run the LanthaScreen TR-FRET ER alpha Coactivator Assay in antagonist mode (to identify antagonist compounds), ER alpha-LBD is added to ligand test compounds, followed by addition of a mixture of agonist, fluorescein-coactivator peptide, and terbium (Tb)-labeled anti-GST antibody. The concentration of agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Co-Factor Interaction Assay, Nuclear Receptor Assay
Product LineLanthaScreen™
Quantity800 x 20 μL assays
TypeCoactivator Assay Kit
Unit Size800 x 20 µL assays
Contents & Storage
1 tube ER alpha Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
1 tube of Peptide (store at -5 to -30°C)
2 bottles TR-FRET Buffer (store at -5 to -30°C)
1 tube Tb-Anti-GST Ab (store at -5 to -30°C)
1 tube of DTT (store at -5 to -30°C)