Invitrogen™

TRA-1-60 Alexa Fluor™ 594 Conjugate Kit for Live Cell Imaging

Catalog number: A24882
Have Questions?
Invitrogen™

TRA-1-60 Alexa Fluor™ 594 Conjugate Kit for Live Cell Imaging

Catalog number: A24882
Have Questions?
Catalog Number
A24882
Unit Size
50 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A2488250 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits are the only kits offering superior imaging for live human PSCs in one box. This TRA-1-60 Alexa Fluor™ 594 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.

With the TRA-1-60 Alexa Fluor™ 594 Conjugate Kit for Live Cell Imaging, you can:
• Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
• Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM imaging medium
• Stain live cell samples with confidence using reagents that are tested to be free of common contaminants

Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM imaging medium.

Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM imaging medium maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.

Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2 μm sterile-filtered and tested to be free of common contaminants.

What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 594 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 uL staining volume and includes:
• 200 μL of Alexa Fluor™ 594 Mouse Anti-Human TRA-1-60 (50X concentration)
• 500 mL FluoroBrite™ DMEM
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
Alexa Fluor 594
Product Type
Conjugate Kit

Contents & Storage

  • 200 μL of Alexa Fluor™ 594 Mouse Anti-Human TRA-1-60 (0.25 mg/mL) (store at 2°C to 8°C)
  • 500 mL FluoroBrite™ DMEM
  • Store at 2°C to 8°C, protect from light

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text