1x1 image pixel for data collection
Ion S5™ Controls Kit Plus
Ion S5™ Controls Kit Plus
Ion Torrent™
Ion S5™ Controls Kit Plus
The Ion S5™ Controls Kit Plus provides library preparation and templating controls for use with the Ion S5™ and IonRead more
Have Questions?
Catalog NumberQuantity
A307291 kit
Catalog number A30729
Price (USD)
1,300.00
Each
Add to cart
Quantity:
1 kit
Price (USD)
1,300.00
Each
Add to cart
The Ion S5™ Controls Kit Plus provides library preparation and templating controls for use with the Ion S5™ and Ion S5™ XL Sequencing Systems. The kit offers greater experimental control and empowers you to assess sample quality, help save time and reagents, and maximize productivity. Each kit contains a CEPH genomic DNA control for up to 3 library preparations, a 200 bp human CEPH control library, and a 400 bp E.Coli control library, for up to 10 template preparations each. The Ion S5 Controls Kit Plus replaces the previous Ion S5™ Controls Kit (Cat. No. A27760).
Note:
This kit is only compatible with the Ion S5 and Ion S5 XL Sequencing Systems.
Specifications
For Use With (Equipment)Ion GeneStudio S5 System, Ion S5™ XL System
Product TypeControls Kit Plus
Quantity1 kit
Product LineIon Torrent™
Unit SizeEach
Frequently asked questions (FAQs)
I would like to perform a template run with Human CEPH Control 200 Library using the Ion 540 Kit-Chef. In the Ion Torrent Browser where I set up the plan, the library selection pulldown menu does not have human CEPH as an option. What should I do?
The definition of the library kit in the plan run is optional. At the moment, there is an option for the CEPH library control and you can leave that field blank. The two parameters that are important for the control libraries are: the correct amount of flows (500 for 200 bp libraries or 850 for 400 bp libraries) and the correct reference (hg19 for CEPH and E. coli DH10B for E. coli).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Share catalog number, name or link