Invitrogen™
Antibody Conjugate Purification Kit, 50-100 μg conjugate
Catalog number: A33088
Have Questions?
Invitrogen™
Antibody Conjugate Purification Kit, 50-100 μg conjugate
Catalog number: A33088
Have Questions?
Catalog Number
A33088
Unit Size
5 purifications
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A330885 purifications
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Molecular Probes™ Antibody Conjugate Purification kits efficiently purify antibody conjugates from unconjugated dye or biotin. This particular kit is optimized for the purification of 50–100 μg of antibody conjugate. Purification is accomplished using an easy-to-assemble size-exclusion spin column. Each kit contains sufficient material for 5 separate purifications, and can also be used to purify high-molecular weight proteins (> 30 kDa) away from unconjugated labels.
Additional Features of this Antibody Conjugate Purification Kit:
• Purified conjugate ready-to-use in ∼45 minutes with ∼15 minutes of hands-on time
• Designed for low-molecular weight labels (less than 3000 Da, e.g. Alexa Fluor™ dyes); cannot be used for removing unconjugated high-molecular weight labels (>10 kDa) such as enzymes (HRP) or proteins (R-phycoerythrin)
More about Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental set-up. To learn more about our labeling kits, see Kits for Labeling Proteins and Nucleic Acids in The Molecular Probes™ Handbook or visit our antibody labeling page.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
Antibody Conjugate Purification Kit
Format
Spin Column
Contents & Storage
Store in refrigerator (2-8 °C).