Applied Biosystems™

SeqStudio™ Cartridge

Catalog number: A33671
Have Questions?
Applied Biosystems™

SeqStudio™ Cartridge

Catalog number: A33671
Have Questions?
Catalog Number
A33671
Unit Size
125 injections/500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A33671125 injections/500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems SeqStudio Cartridge is an all-in-one and easy-to-use reagent cartridge that includes capillaries, universal polymer, buffer, and pump required to run on the SeqStudio Genetic Analyzer System. The universal polymer allows for flexibility to perform Sanger sequencing and fragment analysis on the same instrument run with the same cartridge. Just load your samples, click in the cartridge, and go.

Key features:
• Minimize hands-on setup time with all-in-one cartridge
• Track consumables usage through radio frequency identification (RFID)
• Extended on-instrument 4-month shelf life for cartridge after opening
• Allows for 125 injections or 500 reactions per cartridge
• Responsible, simple, no-charge recycling program

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capillary Length
28 cm
For Use With (Application)
Rare Variant Sequencing, Resequencing, Comparative Sequencing (Somatic Mutations - 10:50), Fragment Analysis (Denaturing), Short Read Sequencing, De Novo Sequencing - Low to Mid Throughput, Short PCR Product Sequencing
Product Line
SeqStudio
Type
Reagent Cartridge
Green Features
Sustainable disposal
Shipping Condition
Wet Ice

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text