Invitrogen™

Collibri™ Library Amplification Master Mix

Catalog number: A38539050
Have Questions?
Invitrogen™

Collibri™ Library Amplification Master Mix

Catalog number: A38539050
Have Questions?
Catalog Number
A38539050
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3853905050 reactions
-
A38539250250 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Invitrogen Collibri Library Amplification Master Mix is a ready-to-use solution designed for amplification of next-generation sequencing (NGS) libraries. The master mix includes Platinum SuperFi DNA Polymerase in combination with a proprietary reaction buffer containing all necessary components for efficient library amplification with minimal bias and superior accuracy.

Features include:
High fidelity—minimal error rates similar to PCR-free methods
High efficiency—high yields from minute input amounts
Minimal bias—uniform amplification regardless of GC content

Platinum SuperFi DNA Polymerase has both 5’ to 3’ polymerase and 3’ to 5’ exonuclease (proofreading) activities, but lacks 5’ to 3’ exonuclease activity. Exceptionally strong proofreading activity ensures amplification of NGS libraries with supreme sequence accuracy, producing NGS error rates similar to PCR-free methods.

The proprietary reaction buffer contains all necessary PCR components and additives to enable efficient and uniform library amplification regardless of GC content, helping improve coverage across GC- and AT-rich sequences and other complex regions. High amplification efficiency enables the use of fewer cycles to achieve the desired yield.

Platinum hot-start technology inhibits DNA polymerase activity at ambient temperatures, allowing room temperature reaction setup and storage of pre-assembled PCR reactions for up to 24 hours prior to the PCR.

The Collibri Library Amplification Master Mix is supplemented with an inert blue dye that provides a visual aid when pipetting and decreases the risk of pipetting errors during reaction setup.

For increased convenience, we offer Collibri Library Amplification Master Mix with Primer Mix, where the master mix is supplied with a 10X Primer Mix that targets the P5 and P7 regions of Illumina adapters and that is supplemented with an inert yellow dye. Mixing both components in a PCR reaction turns the final PCR solution green, further decreasing the risk of pipetting errors.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
DNA, RNA
Fidelity (vs. Taq)
≥100
GC-Rich PCR Performance
High
For Use With (Application)
Sequencing
Hot Start
Built-In Hot Start
No. of Reactions
50
Overhang
Blunt
Product Line
Collibri™
Product Type
Library Amplification Master Mix
Reaction Format
SuperMix or Master Mix
Shipping Condition
Dry Ice
Test Time
Standard
Form
Liquid

Contents & Storage

1 x 1.25 mL Collibri Library Amplification Master Mix (2X)

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text