Invitrogen™

Pre-miR™ miRNA Precursor Molecules—Negative Control #2

Catalog number: AM17111
Have Questions?
Invitrogen™

Pre-miR™ miRNA Precursor Molecules—Negative Control #2

Catalog number: AM17111
Have Questions?
Catalog Number
AM17111
Unit Size
5 nmol
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM171115 nmol
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
A synthetic miRNA mimic for miRNA functional analysis. 5 nmol is supplied.

• Optimized design results in correct strand utilization by RISC
• Purified and ready to transfect

Ambion™ Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments. Pre-miR™ Negative Control #2 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Tube
Control Type
Negative Control
RNAi Type
miRNA Mimic
Species
Human, Mouse, Rat
For Use With (Application)
miRNA & Non-Coding RNA Analysis
Label or Dye
Unconjugated
Product Line
Ambion™, Pre-miR™
Product Type
miRNA Mimic
Quantity
5 nmol
Shipping Condition
Room Temperature

Contents & Storage

Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text