SDS, 10% Solution, RNase-free
Inquire about OEM or Commercial Supply version of this product here.
SDS, 10% Solution, RNase-free
Invitrogen™

SDS, 10% Solution, RNase-free

Molecular biology grade, Ambion SDS, 10% Solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM98231 L
AM9822500 mL
Catalog number AM9823
Price (USD)/ Each
178.65
Online exclusive
190.00 
Save 11.35 (6%)
-
Add to cart
Quantity:
1 L
Price (USD)/ Each
178.65
Online exclusive
190.00 
Save 11.35 (6%)
Add to cart
SDS, 10% Solution, RNase-free
Catalog numberAM9823
Price (USD)/ Each
178.65
Online exclusive
190.00 
Save 11.35 (6%)
-
Add to cart

Molecular biology grade, Ambion SDS, 10% Solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. SDS is used as a component of both hybridization and wash buffers for northern and Southern blots and has many other uses around the lab.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.1
Packaging TypeBottle
Product TypeSDS
Product LineAmbion™
Quantity1 L
Shipping ConditionRoom Temperature
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.