Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Quantity | 2 Vials (1 Green, 1 Red) |
Excitation/Emission | ∼590/610 nm, ∼502/530 nm |
Color | Red, Green |
Detection Method | Fluorescence |
Conjugate | Nuclear Dye |
Form | Powder |
Unit Size | 2 vials |
Quantity | 1 Vial |
Excitation/Emission | ∼590/610 nm |
Color | Red |
Detection Method | Fluorescence |
Conjugate | Nuclear Dye |
Form | Powder |
Unit Size | 1 vial |
Quantity | 5 Vials |
Excitation/Emission | ∼590/610 nm |
Color | Red |
Detection Method | Fluorescence |
Conjugate | Nuclear Dye |
Form | Powder |
Unit Size | 5 vials |
Quantity | 1 Vial |
Excitation/Emission | ∼502/530 nm |
Color | Green |
Detection Method | Fluorescence |
Conjugate | Nuclear Dye |
Form | Powder |
Unit Size | 1 vial |
Quantity | 5 Vials |
Excitation/Emission | ∼502/530 nm |
Color | Green |
Detection Method | Fluorescence |
Conjugate | Nuclear Dye |
Form | Powder |
Unit Size | 5 vials |
Quantity | 1 x 100 μL Vial |
Product Type | Caspase 3/7 Green Detection Reagent |
Excitation/Emission | ∼502/530 nm |
Color | Green |
Shipping Condition | Room Temperature |
Incubation Time | 30 min |
Reaction Time | 30 min |
Product Line | CellEvent™ |
Detection Method | Fluorescence |
Conjugate | CellEvent Caspase 3/7 Green |
For Use With (Equipment) | Fluorescence Microscope, High Content Instrument |
Label Type | Other Label(s) or Dye(s) |
Unit Size | 100 µL |
Quantity | 1 x 25 μL Vial |
Product Type | Caspase 3/7 Reagent |
Excitation/Emission | ∼502/530 nm |
Color | Green |
Shipping Condition | Room Temperature |
Product Line | CellEvent™ |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope, High Content Instrument |
Label or Dye | CellEvent™ Caspase 3/7 Green |
Label Type | Other Label(s) or Dye(s) |
Form | 2 mM in DMSO |
Unit Size | 25 µL |
Catalog Number | Specifications | Unit Size | Color | Excitation/Emission | Quantity | dimFormat | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C10434 | 2 vials | Red, Green | ∼590/610 nm, ∼502/530 nm | 2 Vials (1 Green, 1 Red) | Price: 540.00 Online price: 501.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10430 | 1 vial | Red | ∼590/610 nm | 1 Vial | Price: 270.00 Online price: 250.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10431 | 5 vials | Red | ∼590/610 nm | 5 Vials | Price: 780.00 Online price: 723.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10432 | 1 vial | Green | ∼502/530 nm | 1 Vial | Price: 270.00 Online price: 250.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10433 | 5 vials | Green | ∼502/530 nm | 5 Vials | Price: 780.00 Online price: 723.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10423 | 100 µL | Green | ∼502/530 nm | 1 x 100 μL Vial | Price: 762.00 Online price: 706.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10723 | 25 µL | Green | ∼502/530 nm | 1 x 25 μL Vial | Price: 278.00 Online price: 257.65 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|