1x1 image pixel for data collection
ELISA Phosphate Coating Buffer
ELISA Phosphate Coating Buffer
Thermo Scientific™

ELISA Phosphate Coating Buffer

ELISA Phosphate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developingRead more
Have Questions?
Catalog NumberQuantity
CB07100100 mL
Catalog number CB07100
Price (USD)
-
Quantity:
100 mL
ELISA Phosphate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA. The buffer is a 10 mM phosphate buffer, pH 7.4.

For a complete kit of ELISA buffers, see ELISA Buffer Kit.
View all available buffers and reagents for ELISA applications.

Specifications
ConjugateEnzyme Labeled
For Use With (Application)ELISA
FormConcentrate
Product TypeCoating Buffer
Quantity100 mL
Unit SizeEach
Contents & Storage
Buffer. Store at 2°C to 8°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link