Invitrogen™

ER-Tracker™ Blue-White DPX, for live-cell imaging

Catalog number: E12353
Have Questions?
Invitrogen™

ER-Tracker™ Blue-White DPX, for live-cell imaging

Catalog number: E12353
Have Questions?
Catalog Number
E12353
Unit Size
20 x 50 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
E1235320 x 50 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
ER-Tracker Blue-White DPX dye is a photostable probe that is selective for the endoplasmic reticulum (ER) in live cells. The cells can be aldehyde-fixed after staining with the dye but significant fluorescence signal will be lost. With excitation at ∼374 nm, the environmentally-sensitive probe exhibits emission between 430 and 640 nm. A long-pass DAPI filter works best.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Blue
Concentration
1 mM
For Use With (Equipment)
Fluorescence Microscope
Detection Method
Fluorescent
Product Line
ER-Tracker™
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Dye
SubCellular Localization
Endoplasmic Reticulum

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text