Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Image-iT LIVE Red Poly Caspases Detection Kit |
Format | Slide |
Product Type | Caspase Assay Kit |
Excitation/Emission | SR-VAD-FMK: 550/595 SYTOX Green: 504/523 Hoechst 33342: 350/461 |
No. of Reactions | 25 tests (labeling volumes of 300μL) |
Color | Red, Green, Blue |
Shipping Condition | Room Temperature |
Product Line | Image-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | SR-VAD-FMK, SYTOX Green, Hoechst 33342 |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Enzyme | Poly Caspases |
Unit Size | 1 kit |
Description | Image-iT LIVE Red Caspase-3 and -7 Detection Kit |
Format | Slide |
Product Type | Caspase 3/7 Reagent |
Excitation/Emission | SR-DEVD-FMK: 550/595 SYTOX Green: 504/523 Hoechst 33342: 350/461 |
No. of Reactions | 25 tests (labeling volumes of 300μL) |
Color | Red, Green, Blue |
Shipping Condition | Room Temperature |
Product Line | Image-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | SR-DEVD-FMK, SYTOX Green, Hoechst 33342 |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Enzyme | Caspase 3/7 |
Unit Size | 1 kit |
Description | Image-iT LIVE Green Poly Caspases Detection Kit |
Format | Slide |
Product Type | Caspase Assay Kit |
Excitation/Emission | FAM-VAD-FMK: 488/530 propidium iodide (PI): 535/617 Hoechst 33342: 350/461 |
No. of Reactions | 25 tests (labeling volumes of 300μL) |
Color | Green, Red, Blue |
Shipping Condition | Room Temperature |
Product Line | Image-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | FAM-VAD-FMK, propidium iodide (PI), Hoechst 33342 |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Enzyme | Poly Caspases |
Unit Size | 1 kit |
Description | Image-iT LIVE Green Caspase-3 and -7 Detection Kit |
Format | Slide |
Product Type | Caspase 3/7 Reagent |
Excitation/Emission | FAM-DEVD-FMK: 488/530 propidium iodide (PI): 535/617 Hoechst 33342: 350/461 |
No. of Reactions | 25 tests (labeling volumes of 300μL) |
Color | Green, Red, Blue |
Shipping Condition | Room Temperature |
Product Line | Image-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | FAM-DEVD-FMK, propidium iodide (PI), Hoechst 33342 |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Enzyme | Caspase 3/7 |
Unit Size | 1 kit |
Catalog Number | Specifications | Unit Size | Color | Enzyme | Label or Dye | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
I35101 | 1 kit | Red, Green, Blue | Poly Caspases | SR-VAD-FMK, SYTOX Green, Hoechst 33342 | Price: 470.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
I35102 | 1 kit | Red, Green, Blue | Caspase 3/7 | SR-DEVD-FMK, SYTOX Green, Hoechst 33342 | Price: 538.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
I35104 | 1 kit | Green, Red, Blue | Poly Caspases | FAM-VAD-FMK, propidium iodide (PI), Hoechst 33342 | Price: 319.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
I35106 | 1 kit | Green, Red, Blue | Caspase 3/7 | FAM-DEVD-FMK, propidium iodide (PI), Hoechst 33342 | Price: 319.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|