1x1 image pixel for data collection
GRAMS/AI™ Spectroscopy Software
GRAMS/AI™ Spectroscopy Software
Thermo Scientific™
GRAMS/AI™ Spectroscopy Software
Tired of having to use a different software package for every spectroscopy instrument? This software facilitates data access and collaboration and reduces software training costs.
Have Questions?
Catalog NumberFor Use With (Equipment)
INF-15000GRAMS
Catalog number INF-15000
Price (USD)
-
Request A Quote
For Use With (Equipment):
GRAMS
Long-recognized as the industry standard for visualizing, processing and managing spectroscopy data, Thermo Scientific™ GRAMS/AI™ Spectroscopy Software Suite provides a set of complementary, fully integrated applications that increase productivity across all areas of spectroscopic analysis.
The suite's core application, GRAMS/AI, eliminates the need for multiple instrument software packages by providing a single, integrated environment for data management and analysis. It streamlines data access and facilitates scientific collaboration, as well as reducing overall software training costs.
A comprehensive processing, visualization and reporting package, GRAMS/AI includes an extensive library of built-in data processing routines. Together with advanced data comparison and visualization features, the software gives users the tools they need to solve the toughest analytical problems.
GRAMS/AI handles data from practically any type of analytical instrument—including UV-Vis, fluorescence, FTIR, Raman, NIR and NMR—as well as hyphenated techniques such as GC-MS, LC-MS and LC-PDA.
Multiple Instruments and File Formats
  • Automatically recognizes hundreds of different instrument file formats; can also interface directly for full control and data acquisition via a plug-in My Instrument interface
  • Includes capabilities for interchanging data with Microsoft™ Excel™, real time visualization of large data sets using GRAMS/3D, and accessing data from other GRAMS applications such as Spectral DB databases and Spectral ID search libraries
  • Supports popular general-purpose data formats such as SPC, ASCII, JCAMP and AnDI/NetCDF
  • Use of the universal SPC file format for data storage lets scientists share data with colleagues easily and analyze it right at their desks
  • Compatible with data files from hundreds of different instrument control applications, including Agilent/HP, Beckman, Bio-Rad, Bruker, Gilson, Hitachi, PerkinElmer, Shimadzu, Varian, Waters/Micromass and others
Friendly, Familiar Interface
  • Intuitive interface lets users quickly navigate through open data files, elicit important file information and access parameter settings—all with a click of the mouse
  • Makes it easy for users to create customized workspaces for viewing, reporting, and processing their data
Enterprise-Wide Collaboration and Productivity
  • Enables large research organizations to realize increased productivity through common data processing software and organization-wide data access
Full Suite of Complementary Productivity Applications
  • GRAMS Envision™ for easily viewing multi-format instrument data and creating standardized reports
  • GRAMS/3D for visualizing multi-dimensional data sets in real time
  • Spectral DB for creating shared spectral databases
  • Spectral ID and Spectral ID Server for performing fast, efficient qualitative spectral identifications through spectral library searching
  • GRAMS IQ™ for creating qualitative and quantitative chemometric calibration models
  • IQ Predict for deploying GRAMS IQ calibrations
  • MyInstrument and Software Components for boiling GRAMS into your own programming environment
  • QuickQuant for creating simple quantitative calibration models
  • Thermo Scientific™ Remspec ReactionSleuth™ ActiveApp™ for analyzing spectroscopic reaction data
  • ExcelExchange for data interchange with Microsoft™ Excel™ worksheets
  • Active Multifile Tool for creating and manipulating three-dimensional data sets
  • In addition, several third-party vendors offer GRAMS-compatible applications
Specifications
Certifications/ComplianceFDA Guidelines, IEEE Software Engineering Standards
DescriptionGrams/AI
For Use With (Equipment)GRAMS
TypeSpectroscopy Software
Unit SizeEach
Share catalog number, name or link