OXSAS™ Optical Emission Analytical Software
OXSAS™ Optical Emission Analytical Software
OXSAS™ Optical Emission Analytical Software
OXSAS™ Optical Emission Analytical Software
OXSAS™ Optical Emission Analytical Software
Thermo Scientific™
OXSAS™ Optical Emission Analytical Software
Perform high quality analysis for all your metal analysis jobs easily and quickly with a customizable software solution for optical emission spectrometers.
Have Questions?
產品號碼Type
IQLAAHGABUFABXMATXOXSAS Optical Emission Analytical Software
產品號碼 IQLAAHGABUFABXMATX
價格 (TWD)
-
申請報價
Type:
OXSAS Optical Emission Analytical Software
Perform rapid high quality metals analyses with a customizable software solution for optical emission spectrometers. Flexible and powerful Thermo Scientific™ OXSAS™ Optical Emission Analytical Software runs a variety of analyses, including the development of new analytical methods and calibrations as well as rapid high-quality routine elemental analysis. From simple tasks to complex jobs, the OXSAS software platform can perform many operations in a single click and is designed to evolve to meet your needs throughout your instrument's lifetime.
Template-Driven Analysis
  • Simple, comprehensive analysis of unknown samples.
  • Required analytical parameters and results processing details pre-defined in the routine analysis shortcut.
  • One click creation of a batch with all samples requiring analysis.
  • Automatic Method Choice selects the best analytical parameters set without user intervention
Fast Calibration
  • Multiple analyte curve display, instant base curve calculation, templates for editing and measuring of calibration standards.
  • Calibration curve determination using multi-variable regression with additive and multiplicative correction models.
Familiar User Interface
  • Comprehensive post-treatment of results is available, including export and Microsoft™ Excel support.
  • Integrated Microsoft SQL Server Express relational database stores set up and analyses data.
  • Export and import facilities allow calibration standards, methods or grades to be easily transferred from one instrument to another.
  • Full network and serial transmission capabilities.
  • Common software platform with the WDXRF Thermo Scientific series instrument range.
  • Supports the Thermo Scientific SMS system for full automation of the sample analysis.
  • Monitoring and maintenance tools maximize the system uptime.
  • Measurement Uncertainty and Conformance Probability on unknown samples analysis.
  • SCT Manager: The SCT sample status and history give the overview of the analytical capability of the instrument and of each method any time.
  • Comprehensive, on-line contextual Help.
  • Real, on-line integrated SPC.
  • Key to Metals comprehensive steel and nonferrous metals and cross-reference database included.
Supports the Spark option of the Thermo Scientific™ ARL iSpark™ Plus, the ARL iSpark™ and of the Thermo Scientific™ ARL 4460™ Optical Emission Spectrometer.
Allows complex calculations such as element soluble/insoluble or elements inclusions (number, size and chemical composition) with no additional analysis time.
Options for OXSAS Software
  • Charge correction feature allows the calculation of the weight of materials to be added to a furnace to bring an out-of-specification charge within limits.
  • SPC-Full: On-line Statistical Process Control, Full Graphical Package Extension.
  • ARLcom: Software package for the transmission of results using network and serial communication via Local Area Network to computer applications and printers using TCP/IP or to files.
  • Remote Sample Definition: Allows samples registration from an external application which can be located anywhere on the network.
Compatible with:
ARL iSpark Plus Optical Emission Spectrometer
ARL iSpark Plus Fire Assay Analyzer
ARL iSpark Optical Emission Spectrometer
ARL 4460 Optical Emission Spectrometer
Thermo Scientific™ ARL™ Fire Assay Analyzer
規格
TypeOXSAS Optical Emission Analytical Software
Unit SizeEach
文件與下載
證書
安全資料表
分享目錄編號、產品名稱或連結。