Invitrogen™
Zero Blunt™ PCR Cloning Kit
Catalog number: K270040
Have Questions?
Invitrogen™
Zero Blunt™ PCR Cloning Kit
Catalog number: K270040
Have Questions?
Catalog Number
K270040
Unit Size
40 rxns
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K27004040 rxns
-
K27002020 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Zero Blunt™ PCR Cloning Kit offers an easy method for high-efficiency (>80%) cloning of blunt-end PCR products amplified with proof-reading, thermostable DNA polymerases. The Zero Blunt™ PCR Cloning Kit uses the multipurpose cloning vector pCR™-Blunt and ExpressLink™ T4 DNA Ligase to generate a ligation product in a five-minute, room-temperature ligation step.
Features of the Zero Blunt™ Cloning™ Kit with pCR™-Blunt vector:
Fast & convenient—5-minute, room-temperature ligation
EfficientccdB gene for positive selection results in >80% clones with correct insert
Flexible—choice of kanamycin or Zeocin™ resistance for flexible antibiotic selection
The pCR™-Blunt vector provides:
EcoR I sites flanking the PCR product insertion site for excision of inserts
• T7 promoter/primer site for in vitro RNA transcription and sequencing
• M13 forward and reverse primer sites for sequencing or PCR screening
How Zero Blunt™ PCR Cloning Works
The Zero Blunt™ PCR Cloning Kit is designed to clone blunt PCR fragments (or any blunt DNA fragment) with a low background of non-recombinants. The pCR™-Blunt vector contains the lethal E. coliccdB gene fused to the C-terminus of LacZα (Bernard et al., 1994). Ligation of a blunt PCR fragment disrupts expression of the lacZα-ccdB gene fusion permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed when the transformation mixture is plated.
Kit Configurations
The Zero Blunt™ PCR Cloning Kit is offered in a variety of configurations: with One Shot™ TOPO10 Chemically Competent E. coli (K2700-20 and K2700-40) and without competent cells (K2750-20 and K2750-40) in 20- and 40- reaction kit sizes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast Strain
TOP10
Cloning Method
Blunt PCR
Format
Kit
Product Line
Zero Blunt™
Product Type
PCR Cloning Kit
Promoter
T7
Quantity
40 Reactions
Vector
Blunt DNA Cloning Vectors
Unit Size
40 rxns
Contents & Storage
Zero Blunt™ PCR cloning kits contain linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
Store the One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.