Invitrogen™

PVDF/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm

Catalog number: LC2002
Have Questions?
Invitrogen™

PVDF/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm

Catalog number: LC2002
Have Questions?
Catalog Number
LC2002
Unit Size
20 membrane/filter paper sandwiches
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC200220 membrane/filter paper sandwiches
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
PVDF/Filter Paper Sandwiches, 0.2 μm, have excellent binding properties for western blotting, dot-blot assays, and other protein or nucleic acid methods such as protein sequencing. The 0.2-μm pore size makes these membranes ideal for protein analysis of small amounts of proteins (as little as 10 pmoles), amino acid analysis, and transfer of small molecular weight proteins and peptides.

See all membranes and filter papers ›

Features
Membrane: polyvinylidene difluoride (PVDF)
Filter paper thickness: 0.8 mm
Binding capacity: 50–150 μg/cm2 for large, globular proteins; >150 μg/cm2 for smaller peptides
Re-probe characteristics: yes
Pre-activation: required with 100% alcohol (methanol, ethanol, or isopropanol)
Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
Durability: compatible with most organic solvents, acids, and mild bases
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Length (Metric)
8.3 cm
Width (Metric)
7.3 cm
Format
Sandwich
Material
PVDF
Pore Size
0.2 μm
Shipping Condition
Room Temperature
Dimensions (LxW)
7.3 cm x 8.3 cm
Thickness
0.8 mm
Product Line
Novex

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text