Invitrogen™

Blotting Roller

Catalog number: LC2100
Have Questions?
Invitrogen™

Blotting Roller

Catalog number: LC2100
Have Questions?
Catalog Number
LC2100
Unit Size
1 unit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC21001 unit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Blotting Roller is an 8.6 cm wide roller designed for easy and efficient removal of bubbles when assembling gel/membrane sandwiches for blotting both E-PAGE™ gels and mini-gels such as NuPAGE® and Novex® Tris-Glycine gels. It consists of a Delrin roller attached to a stainless steel handle.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Western Blot
Gel Compatibility
E-PAGE™ Gel, Novex™ Mini Gel, NuPAGE™ Gel
Product Line
Novex
Type
Blotting Roller
Shipping Condition
Room Temperature

Contents & Storage

One blotting roller.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text