Invitrogen™

Bolt™ Empty Mini Gel Cassettes

Catalog number: NW2010
Have Questions?
Invitrogen™

Bolt™ Empty Mini Gel Cassettes

Catalog number: NW2010
Have Questions?
Catalog Number
NW2010
Unit Size
20 cassettes
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
NW201020 cassettes
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Bolt Empty Gel Cassettes give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank and XCell SureLock Mini-Cell. The cassettes are sealed on three sides (so no gel leakage) and feature the unique wedge-shaped well. They are available in the standard 1-mm thickness and can be used with Bolt™ 10- or 12-well combs (Cat. Nos. NW3010 & NW3012). Since Bolt™ empty cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mini Gel Tank and XCell SureLock Mini-Cell
Gel Size
Mini (8 x 8 cm)
Product Line
Bolt
Type
Empty Mini Gel Cassette
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text