Informational:Informational:New, second-generation pHrodo dyes are more soluble and reactive - ideal for antibody labeling.
Invitrogen™

pHrodo™ Green STP Ester

Catalog number: P35369
Have Questions?
Invitrogen™

pHrodo™ Green STP Ester

Catalog number: P35369
Have Questions?
Catalog Number
P35369
Unit Size
500 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P35369500 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The amine-reactive pH-sensitive pHrodo™ Green STP ester dye is suitable for the creation of bioconjugates to study endocytosis and phagocytosis. pHrodo™ Green dramatically increases fluorescence as the pH of its surroundings become more acidic.
• Use pH-sensitive pHrodo™ Green STP ester to make pH-sensitive bioconjugates of your choice
• Get faster, more accurate results than with any other endocytosis or phagocytosis assay—no need for wash steps or quenchers
• Multiplex with red fluorescent markers such as RFP, pHrodo™ Red, and many others

The increase in fluorescence of pHrodo™ Green as pH changes from basic to acidic correlates with the acidification of intracellular vesicles, making it an ideal tool to study endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo™ Green makes it useful for multi-color experiments. pHrodo™ Green can be detected using most standard green fluorescent filter sets and has been validated for use on a variety of platforms, including flow cytometry, fluorescent microscopy, and high content screening (HCS). The lack of fluorescence of pHrodo™ Green in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.

The pHrodo™ Green STP ester is an amine reactive dye that can be used to make pHrodo™ Green bioconjugates in aqueous buffer. The STP ester will react with primary amines on a protein, cell, or virus to create a stable conjugate that can be used in live cell assays or stored for later use.

Amine reactive pHrodo™ dye is also available with red fluorescence (see pHrodo™ Red SE). In addition, thiol-reactive pHrodo™ Red Maleimide and Green Maleimide are a great alternative to amine-reactive dyes for antibody labeling. Thiol labeling ensures that the dye will not attach to the binding region of the antibody. pHrodo™ Green and Red conjugates, for example dextran, E. coli and S. aureus, are also available in ready-to-use form.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
500 μg
Product Type
STP Ester
Solubility
DMSO (Dimethylsulfoxide)
Chemical Reactivity
Amine
Detection Method
Fluorescent
EC Extinction Coefficient
75000
Reactive Group
STP Ester
Shipping Condition
Room Temperature
Label or Dye
pHrodo™ Green
Color
Green
Product Line
pHrodo™
Emission
525
Excitation
505

Contents & Storage

1 vial , store ≤ -20 °C, dessicate and protected from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text