Invitrogen™

AccuCheck Counting Beads

Catalog number: PCB100
Have Questions?
Invitrogen™

AccuCheck Counting Beads

Catalog number: PCB100
Have Questions?
Catalog Number
PCB100
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PCB10010 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
AccuCheck Counting Beads are an efficient single-platform method for absolute cell counts that combines the advantages of direct flow cytometric immunophenotyping with the use of two different fluorescent beads (A and B beads). These two fluorospheres are used as a double internal standard for blood volume calculation. A known volume of AccuCheck Counting Beads is added to the same known volume of stained blood in a lyse-no-wash technique. The beads are counted along with cells. Since the concentration of beads is known, the number of cells per microliter (the absolute count) is obtained by relating the number of cells counted to the total number of fluorescent bead events. The cell number is then multiplied by the number of total fluorospheres per unit of volume. As the AccuCheck Counting Beads system contains two different fluorospheres in a known proportion, we can first assure the accuracy of the assay by verifying the proportion of both types of beads.

View information about all flow cytometry cell counting beads.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
Diameter (Metric)
6.36 μm, 6.4 μm
Emission
488 and 633/635
Excitation Wavelength Range
Bead A: 488/575-585 nm, Bead B: 635/660-680 nm
For Use With (Equipment)
Flow Cytometer
Format
Solution
No. of Tests
100
Shipping Condition
Wet Ice
Product Type
Cell Counting Bead

Contents & Storage

Contains 1 bottle of AccuCheck counting beads (10 mL). Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text