The Phosphate Sensor is a highly sensitive reagent for the detection of inorganic phosphate, either in real-time kinetic or end-pointRead more
Have Questions?
Change view
Catalog Number
Quantity
PV4407
100 nmol; 10,000 assay(s)
PV4406
10 nmol
2 Options
Catalog number PV4407
Price (USD)/ Each
2,663.65
Online exclusive
2,860.00
Save 196.35 (7%)
-
Add to cart
Quantity:
100 nmol; 10,000 assay(s)
Price (USD)/ Each
2,663.65
Online exclusive
2,860.00
Save 196.35 (7%)
Add to cart
Phosphate Sensor
Catalog numberPV4407
Price (USD)/ Each
2,663.65
Online exclusive
2,860.00
Save 196.35 (7%)
-
Add to cart
The Phosphate Sensor is a highly sensitive reagent for the detection of inorganic phosphate, either in real-time kinetic or end-point mode. This product is a purified form of recombinant E. coli phosphate-binding protein labeled with the fluorophore MDCC, which is sensitive to changes in its environment. Binding of inorganic phosphate by Phosphate Sensor is rapid and tight (Kd ∼ 0.1 μM), resulting in a large increase in fluorescence. The Phosphate Sensor detects phosphate in the high-nanomolar to low-micromolar range, which is 100-fold more sensitive than malachite green. This reagent provides you with a sensitive assay for detecting enzymes that produce inorganic phosphate either directly or indirectly through coupled reactions, such as phosphatases, ATPases, GTPases, and phosphodiesterases.
For Research Use Only. Not for use in diagnostic procedures.