Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay | dsDNA Quantitation, High Sensitivity |
Detection Method | Fluorescence |
Excitation/Emission | 510/527 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 500 Reactions |
Quantitation Range | 0.1 to 120 ng |
Product Line | Qubit |
Shipping Condition | Room Temperature |
Unit Size | 500 assays |
Assay | dsDNA Quantitation, Broad Range |
Detection Method | Fluorescence |
Excitation/Emission | 510/527 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 100 Reactions |
Quantitation Range | 4 to 2000 ng |
Product Line | Qubit |
Shipping Condition | Room Temperature |
Unit Size | 100 assays |
Assay | dsDNA Quantitation, Broad Range |
Detection Method | Fluorescence |
Excitation/Emission | 510/527 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 500 Reactions |
Quantitation Range | 4 to 2000 ng |
Product Line | Qubit |
Shipping Condition | Room Temperature |
Unit Size | 500 assays |
Assay | dsDNA Quantitation, High Sensitivity |
Detection Method | Fluorescence |
Excitation/Emission | 510/527 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 100 Reactions |
Quantitation Range | 0.1 to 120 ng |
Product Line | Qubit |
Shipping Condition | Room Temperature |
Unit Size | 100 assays |
Catalog Number | Specifications | Unit Size | Assay | Quantitation Range | No. of Reactions | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q32854 | 500 assays | dsDNA Quantitation, High Sensitivity | 0.1 to 120 ng | 500 Reactions | 366.65 Online price 389.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32850 | 100 assays | dsDNA Quantitation, Broad Range | 4 to 2000 ng | 100 Reactions | 126.65 Online price 131.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32853 | 500 assays | dsDNA Quantitation, Broad Range | 4 to 2000 ng | 500 Reactions | 364.65 Online price 387.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32851 | 100 assays | dsDNA Quantitation, High Sensitivity | 0.1 to 120 ng | 100 Reactions | 126.65 Online price 131.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|