Thermo Scientific™
ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay
Catalog number: R2474048
Have Questions?
Thermo Scientific™
ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay
Catalog number: R2474048
Have Questions?
Rapidly detect Shiga Toxin-producing Escherichia coli in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay.
Catalog Number
R2474048
Unit Size
Each
Description
ProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Quantity
48 Wells/Kit
Price (USD)
Full specifications
DescriptionProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Detectable AnalytesE. coli Shiga Toxin 1 and 2
Sample TypeFecal
TypeE. coli Testing Kit
For Use With (Application)Enzyme Immunoassay Detection
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity48 Wells/Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Quantity
96 Wells/Kit
Price (USD)
Full specifications
DescriptionProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Detectable AnalytesE. coli Shiga Toxin 1 and 2
Sample TypeHuman Fecal
TypeE. coli Testing Kit
For Use With (Application)Enzyme Immunoassay Detection
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity96 Wells/Kit
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionQuantityPrice (USD)
R2474048Full specifications
EachProSpecT STEC (Shiga Toxin E.coli) Microplate Assay48 Wells/KitRequest A Quote
DescriptionProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Detectable AnalytesE. coli Shiga Toxin 1 and 2
Sample TypeFecal
TypeE. coli Testing Kit
For Use With (Application)Enzyme Immunoassay Detection
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity48 Wells/Kit
Unit SizeEach
R2474096Full specifications
EachProSpecT STEC (Shiga Toxin E.coli) Microplate Assay96 Wells/KitRequest A Quote
DescriptionProSpecT STEC (Shiga Toxin E.coli) Microplate Assay
Detectable AnalytesE. coli Shiga Toxin 1 and 2
Sample TypeHuman Fecal
TypeE. coli Testing Kit
For Use With (Application)Enzyme Immunoassay Detection
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity96 Wells/Kit
Unit SizeEach
Showing 2 of 2
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Shiga Toxin-producing Escherichia coli. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT STEC (Shiga Toxin E.coli) is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT STEC provides same-day toxin detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Contents & Storage
2°C to 8°C