Thermo Scientific™

Staphaurex™ Latex Agglutination Test

Catalog number: R30859902
Have Questions?
Thermo Scientific™

Staphaurex™ Latex Agglutination Test

Catalog number: R30859902
Have Questions?
Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, using the Thermo Scientific™ Staphaurex™ Latex Agglutination Test.
 
Catalog Number
R30859902
Unit Size
Each
Description
Staphaurex Latex Agglutination Test
Quantity
400 Tests/Kit
Content And Storage
Price (USD)
Full specifications
Control SetsPositive
DescriptionStaphaurex Latex Agglutination Test
CE MarkerYes
Detectable AnalytesClumping Factor and Protein A
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity400 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Staphaurex Latex Agglutination Test
Quantity
120 Tests/Kit
Content And Storage
Price (USD)
Full specifications
Control SetsPositive
DescriptionStaphaurex Latex Agglutination Test
CE MarkerYes
Detectable AnalytesProtein A
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity120 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionQuantityContent And StoragePrice (USD)
R30859902Full specifications
EachStaphaurex Latex Agglutination Test400 Tests/KitRequest A Quote
Control SetsPositive
DescriptionStaphaurex Latex Agglutination Test
CE MarkerYes
Detectable AnalytesClumping Factor and Protein A
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity400 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
R30859901Full specifications
EachStaphaurex Latex Agglutination Test120 Tests/KitRequest A Quote
Control SetsPositive
DescriptionStaphaurex Latex Agglutination Test
CE MarkerYes
Detectable AnalytesProtein A
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity120 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
Showing 2 of 2
Experience quality and reliability with this rapid and accurate latex test for the identification and differentiation of staphylococcus aureus. White latex particles are coated with human fibrinogen for detection of clumping factor and specific IgG for detection of protein A.

When mixed on a slide with a suspension of S. aureus, reaction of clumping factor with the fibrinogen and/or protein A with the IgG causes rapid, strong agglutination of the latex particles.

Proven Performance
  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with reliable results that have greater than 99% sensitivity and specificity (according to data available in the package inserts)

Workflow Efficiency
  • Clear-cut, robust reactions provide easy-to-read results
  • Improve productivity with compact kit configurations, color-coded reagents, and convenient workstations
  • Flexible use from selective and non-selective media
  • Control latex is not required

Economical Solution
  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Contents & Storage

2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text