Thermo Scientific™
RapID™ YEAST PLUS System
Catalog number: R8311007
Have Questions?
Thermo Scientific™
RapID™ YEAST PLUS System
Catalog number: R8311007
Have Questions?
Identify over 40 different yeasts in just four hours using the Thermo Scientific™ RapID™ YEAST PLUS System, an identification system based on enzyme technology.
Catalog Number
R8311007
Unit Size
Each
Description
RapID YEAST PLUS System
Detectable Analytes
Yeasts
Quantity
20 Panels/Kit
Price (USD)
Full specifications
DescriptionRapID YEAST PLUS System
CE MarkerYes
Detectable AnalytesYeasts
Packaging Type1 Box
Quantity20 Panels/Kit
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionDetectable AnalytesQuantityPrice (USD)
R8311007Full specifications
EachRapID YEAST PLUS SystemYeasts20 Panels/KitRequest A Quote
DescriptionRapID YEAST PLUS System
CE MarkerYes
Detectable AnalytesYeasts
Packaging Type1 Box
Quantity20 Panels/Kit
Unit SizeEach
Showing 1 of 1
The RapID YEAST PLUS System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important yeast, yeast-like and related organisms which have been isolated from human clinical specimens.
  • One-step inoculation—decreased prep time for increased productivity
  • Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
  • Four-hour incubation*—quicker time-to-result for a faster response
  • Common procedure—lessened materials usage saves time and money
  • Visible color reactions—reduced subjectivity decreases repeat testing
  • Better reporting—more coverage, advanced updates
*Two-hour incubation for RapID SS/u.
A clear plastic tray contains reagent impregnated wells, allowing simultaneuos inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Innoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a databse or through the use of a computer-generated Code Compedium (ERIC).
Requires:
RapID Inoculation Fluid (R8325106)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Contents & Storage
2°C to 8°C