CTS™ KnockOut™ DMEM/F-12
CTS™ KnockOut™ DMEM/F-12
Gibco™

CTS™ KnockOut™ DMEM/F-12

CTS™ KnockOut™ DMEM/F-12 CTS™ is a serum free, low osmolality medium without L-glutamine or HEPES buffer that is optimized forRead more
Have Questions?
Catalog number A1370801
Price (USD)/ Each
70.50
-
Add to cart
Quantity:
500 mL
Price (USD)/ Each
70.50
Add to cart
CTS™ KnockOut™ DMEM/F-12
Catalog numberA1370801
Price (USD)/ Each
70.50
-
Add to cart

CTS™ KnockOut™ DMEM/F-12 CTS™ is a serum free, low osmolality medium without L-glutamine or HEPES buffer that is optimized for the growth of human embryonic stem cells (ESC) and induced pluripotent stem cells (iPSC). KnockOut™ DMEM/F-12 CTS™ media is designed for use with KnockOut™ SR and KnockOut™ SR XenoFree CTS™ supplements.

• Designed for reliable and consistent performance
• Perform ESC/iPSC culturing with little to no spontaneous differentiation
• Be ready for clinical research with this cGmP manufactured formula and support documentation

Consistent Results Without Fetal Bovine Serum
Cell Therapy Systems KnockOut™ DMEM/F-12 CTS™ media is part of the KnockOut™ line of media products that have delivered reliable results for ESC and now iPSC culturing for years. This fetal bovine free media leads to little or no spontaneous differentiation, and frees you from repeated screens.

Equivalent Morphology
Stem cells cultured in KnockOut™ DMEM/F-12 CTS™ media show normal morphology and karyotype when compared to control stem cells. You can also expect equivalent morphology to manually passaged hESCs, even after enzymatic passaging.

Simplify Your Transition into Clinical Research
The Gibco™ Cell Therapy Systems (CTS™) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. Gibco™ Cell Therapy Systems (CTS™) products deliver:
• Reduced burden in qualifying reagents
• Included appropriate certificates of analyses and origin
• Convenient access to our Drug Master File (DMF)

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeEmbryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Medium
Shelf Life18 months from date of manufacture
SterilitySterile-filtered
FormatBottle
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity500 mL
Shipping ConditionAmbient
Unit SizeEach
Contents & Storage
Store at 2–8°C and protect from light.