Invitrogen™

SYTOX™ Green Nucleic Acid Stain - 5 mM Solution in DMSO

Catalog number: S7020
Have Questions?
Invitrogen™

SYTOX™ Green Nucleic Acid Stain - 5 mM Solution in DMSO

Catalog number: S7020
Have Questions?
Catalog Number
S7020
Unit Size
250 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S7020250 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SYTOX Green nucleic acid stain is an excellent green-fluorescent nuclear and chromosome counterstain that is impermeant to live cells, making it a useful indicator of dead cells within a population.

Also available as a room-temperature-stable, ready-to-use solution: NucGreen Dead 488 Ready Probes Reagent.
See other Ready Probes ready-to-use imaging reagents and accessories ›

 Impermeant to live cells
 Excitation/Emission: 504/523 nm
 Use with 488 Argon-ion laser
 Works with mammalian cells and both Gram-positive and Gram-negative bacteria

Simple Viability Determination
SYTOX Green allows quick determination of cell viability when using flow cytometers, fluorescence microscopes, fluorometers, or fluorescence microplate readers. It will not cross intact membranes, but will easily penetrate compromised membranes characteristic of dead cells. Because it exhibits >500-fold fluorescence enhancement upon binding nucleic acids, briefly incubating dead cells with SYTOX Green will result in bright green fluorescence with an emission peak of 523 nm when excited by a 488 nm argon-ion laser or any other 450–490 nm source. The exceptional brightness of the signal produced in dead cells makes SYTOX Green particularly useful for determining viability of not just mammalian cells, but both Gram-positive and Gram-negative bacteria.

Part of the Invitrogen Family of Cell Viability Stains and Assays
SYTOX stains for dead cells are available in a variety of colors, including red, blue, and orange. Additionally, Invitrogen has incorporated SYTOX stains into a number of assays for apoptosis, cell viability, and metabolism.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related Link
•  Find out more about all SYTOX Products.



Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Green
Excitation Wavelength Range
504 nm
Dye Type
Cell-Permeant
Format
Tube(s)
For Use With (Equipment)
Flow Cytometer
Volume (Metric)
250 μL
Detection Method
Fluorescence
Emission
523 nm
Form
Solution
Product Line
SYTOX
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Nucleic Acid Stain
SubCellular Localization
Nucleic Acids, Nucleus

Contents & Storage

Store in freezer at -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text