Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACAAGTATGTAAGTGGAACCA[C/T]TGGTGACTCTGGGAGCTGAAGAGCA
Species: |
Human |
dbSNP Submissions: |
NA
|
Phenotype: |
![]() |
Literature Links: |
FSHR PubMed Links |
Allele Nomenclature: |
|
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FSHR - follicle stimulating hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000145.3 | 1832 | Missense Mutation | AAT,AGT | N,S 680 | NP_000136.2 | |
NM_181446.2 | 1832 | Missense Mutation | AAT,AGT | N,S 654 | NP_852111.2 | |
XM_011532733.2 | 1832 | Missense Mutation | AAT,AGT | N,S 714 | XP_011531035.1 | |
XM_011532734.2 | 1832 | Missense Mutation | AAT,AGT | N,S 603 | XP_011531036.1 | |
XM_011532735.2 | 1832 | Missense Mutation | AAT,AGT | N,S 416 | XP_011531037.1 | |
XM_011532736.2 | 1832 | Missense Mutation | AAT,AGT | N,S 416 | XP_011531038.1 | |
XM_011532740.1 | 1832 | Intron | XP_011531042.1 |
Set Membership: |
![]() ![]() |