Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079687.3 | NP_524411.1 | 1 | 1260 | 80 | |
GenBank mRNA | AF184224.1 | - | 1 | 1247 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |