Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142597.4 | NP_650854.1 | 1 | 2677 | 70 | |
GenBank mRNA | AY069370.1 | - | 1 | 2628 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |