Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031535.1 | NP_001026706.1 | 1 - 2 | 702 | 127 | |
XM_015285306.2 | XP_015140792.1 | 2 - 3 | 955 | 127 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |