Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm00839363_m1
            See other G6PC GE Assays ›
            Gene Symbol:
            G6pc
            Gene Name:
            glucose-6-phosphatase, catalytic
            Gene Aliases:
            AW107337, G6Pase, G6pt, Glc-6-Pase
            Chromosome Location:
            Chr.11: 101367730 - 101377903 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            Assay ID Mm00839363_m1
            Size
            Availability Inventoried
            Catalog # 4331182
            Price (USD) 272.00
            Your Price
            Online offer (USD):256.65
            256.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information
            • Citations (2)

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Gene Symbol: G6pc
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_008061.3 NP_032087.2 3 - 4 514 116
            GenBank mRNA AK050279.1 - 3 - 4 529 116
            AK052656.1 - 3 - 4 529 116
            BC013448.1 - 3 - 4 514 4237115 116
            U00445.1 - 3 - 4 529 116

            Back To Top

            More Information


            Set Membership:

            Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene G6pc

            Panther Classification:

            Molecular Function -

            phosphatase

            Gene Ontology Categories:

            Function(s) Process(es)

            glycogen metabolic process
            glycogen catabolic process
            gluconeogenesis
            triglyceride metabolic process
            phosphate-containing compound metabolic process
            steroid metabolic process
            regulation of gene expression
            glucose-6-phosphate transport
            dephosphorylation
            multicellular organism growth
            glucose homeostasis
            cholesterol homeostasis
            urate metabolic process
            phosphorylated carbohydrate dephosphorylation
            glucose 6-phosphate metabolic process
            lipid homeostasis
            catalytic activity
            glucose-6-phosphatase activity
            phosphotransferase activity, alcohol group as acceptor
            hydrolase activity
            phosphate ion binding

            MicroArray Probes:

            Agilent Affymetrix ABI Illumina

            Array Name Probe IDs Targets Target Type
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462385 U00445.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462385 U00445.1 mouse_transcriptome
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334074 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941356 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22117 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90938 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171235 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673649 - -
            Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145156 - -
            Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157154 - -
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462385 U00445.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462385 U00445.1 mouse_transcriptome
            Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462385 U00445.1 mouse_transcriptome
            Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462385 U00445.1 mouse_transcriptome
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073545 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073548 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073547 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073537 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073535 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073546 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073515 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073523 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073554 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073539 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073534 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073524 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073527 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073522 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073519 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073530 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073518 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073551 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073532 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073529 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073544 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073526 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073542 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073517 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145156 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07466499 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157154 - -
            Whole Mouse Genome G4122A A_51_P462385 U00445.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P462385 U00445.1 mouse_transcriptome
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101020 - -
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139061 - -
            Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145156 - -
            Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157154 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09212221 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09313194 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09212216 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09212220 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09212218 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09212217 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073627 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073501 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073558 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073584 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073596 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073499 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073610 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073513 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073561 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073508 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073527 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073554 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073529 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073523 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073573 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073515 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073619 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073588 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073490 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073493 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073577 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073598 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073632 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073567 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073618 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073519 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073616 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073526 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073614 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073591 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073600 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073605 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073603 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073595 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073517 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073511 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073537 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073502 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073510 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073569 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073514 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073544 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073546 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073530 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073623 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073547 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073532 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073628 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073534 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073589 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073494 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073503 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073559 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073578 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073548 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073545 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073582 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073566 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073586 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073592 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073540 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073543 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073534 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073539 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073513 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073572 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073503 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09172215 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073547 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073542 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073523 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073573 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073524 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073551 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073561 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073530 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09723765 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09619686 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09264824 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09701104 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073518 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073556 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073510 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09338548 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073559 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073536 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073514 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073502 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073504 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073564 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09212216 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073537 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073569 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09212221 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09212220 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09479201 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073535 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09479196 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09052323 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073519 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073555 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073511 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09268291 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073570 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073529 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09172206 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073499 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073566 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073571 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073538 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073532 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073548 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P09191879 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073515 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P28073501 - -
            Array Name Probe IDs Targets Target Type
            Affymetrix Mouse Gene 1.0 ST Array 10381391 - -
            Affymetrix Mouse Gene 1.0 ST Array 10381393 - -
            Affymetrix Mouse Gene 1.0 ST Array 10381388 - -
            Affymetrix Mouse Gene 1.0 ST Array 10381390 - -
            Affymetrix Mouse Gene 1.0 ST Array 10381394 - -
            Affymetrix Mouse Gene 1.0 ST Array 10381389 - -
            Affymetrix HT MG-430 PM Array Plate 1417880_PM_at - -
            Affymetrix Mouse Gene 1.1 ST Array 10381387 - -
            Mouse Genome 430 2.0 Array 1417880_PM_at - -
            Mouse Genome 430 2.0 Array 1417880_at - -
            Affymetrix GeneChip Mouse Genome 430 2.0 Array 14377_PM_at - -
            Affymetrix GeneChip Mouse Genome 430 2.0 Array 14377_at - -
            Affymetrix Murine Genome 6500A Array Msa1190_at - -
            Mouse Expression Set 430 1417880_at - -
            Mouse Expression Set 430 1417880_PM_at - -
            Murine Genome U74v2 Set 103333_PM_at - -
            Murine Genome U74v2 Set 103333_at - -
            Mouse Genome 430A 2.0 Array 1417880_at - -
            Mouse Genome 430A 2.0 Array 1417880_PM_at - -
            Affymetrix HT Mouse Genome 430A Array 1417880_PM_at - -
            Affymetrix HT Mouse Genome 430A Array 1417880_at - -
            Array Name Probe IDs Targets Target Type
            ABI Mouse Genome Survey Microarray 333144 - -
            Array Name Probe IDs Targets Target Type
            MouseWG-6 v2 Expression BeadChip 430093 - -
            Illumina mouse-6 v1.0 expression beadchip 430093 - -
            Illumina mouseRef-8 v1.0 expression beadchip 0000430093 - -
            Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245103 - -
            Illumina mouse-6 v1.1 expression beadchip ILMN_1245103 - -
            mouse-6 v1.1 (Illumina) scl40883.5_194-S - -
            Illumina Sentrix mouse-6.v1 bead array 430093 - -
            Illumina Sentrix MouseRef-8 Expression BeadChip scl40883.5_194-S - -
            Illumina Sentrix Mouse-6 Expression BeadChip (2) 430093 - -
            MouseRef-8 v2 Expression BeadChip ILMN_1245103 - -
            Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245103 - -
            Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40883.5_194-S - -

            Back To Top
             

            Citations (2)

            Specific reduction of hepatic glucose 6-phosphate transporter-1 ameliorates diabetes while avoiding complications of glycogen storage disease.
            Authors
            Sloop KW, Showalter AD, Cox AL, Cao JX, Siesky AM, Zhang HY, Irizarry AR, Murray SF, Booten SL, Finger EA, McKay RA, Monia BP, Bhanot S, Michael MD
            Journal
            J Biol Chem - Page No 19113 (Fri Jun 29 00:00:00 EDT 2007)
            Volume (Year)
            282 (2007)
            Product Usage
            -
            Blood glucose-lowering nuclear receptor agonists only partially normalize hepatic gene expression in db/db mice.
            Authors
            Loffler M, Bilban M, Reimers M, Waldhausl W, Stulnig TM
            Journal
            J Pharmacol Exp Ther - Page No 797 (Wed Feb 01 00:00:00 EST 2006)
            Volume (Year)
            316 (2006)
            Product Usage
            -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-s5pk6:80/100.66.78.226:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline