Desktop SEM
Since the introduction of electron microscopes in the 1930s, scanning electron microscopy (SEM) has developed into a critical tool within numerous different research fields, spanning everything from materials science, to forensics, to industrial manufacturing, and even to the life sciences.
As soon as microscopic information about the surface or near-surface region of a specimen is needed, SEM becomes a necessary tool. For that reason, the method finds applications in nearly every branch of science, technology and industry.
Desktop SEM instruments boast enhanced ease of use, democratizing SEM technology. Additionally, they have the added benefit of a reduced frame size. While traditional “floor model” SEM instrumentation can require a specialized room or facility, desktop instrumentation is far more robust. Some have even taken their desktop SEMs on the road, providing on-site electron microscopy analysis via mobile laboratories.
PoroMetric
Desktop SEM for materials science
The interdisciplinary field of materials science focuses on the diverse properties of matter and their potential applications in science and engineering. For that, a thorough analysis of the material is crucial, giving engineers and materials scientists the information they need to create novel materials with desirable properties.
A scanning electron microscope can help you observe and analyze material surfaces and interfaces, either for quality assurance purposes during production or for the analysis of compounds after processing.
The Phenom Desktop SEM product line features versatile and reliable topographic and elemental analysis (via integrated energy-dispersive X-ray spectroscopy (EDS) detectors) combined with an intuitive, easy-to-use software interface that allows even novices to quickly obtain their first high-quality results. This flexibility is ideal for materials science applications, where researchers of varied scientific backgrounds can all obtain vital analytical information.
Style Sheet for Fonts
Style Sheet for Instrument Cards Original
Style Sheet for Komodo Tabs
Technology
Technology
Industry
Applications
Life Sciences
Materials Science
Semiconductors
Techniques
Life Sciences
Materials Science
Semiconductors
Phenom G6 Desktop SEMs
  • Durable high-brightness source
  • Compact design
  • Three models to suit your needs
Helios 6 HXS FIB-SEM
  • Automated material handling
  • Automated data connectivity
  • Automated FIB-SEM workflows
Helios 6 HX FIB-SEM
  • Fully auto TEM sample prep
  • Sub-nm targeting accuracy
  • Robust FIB-SEM workflow
Apreo ChemiSEM System
  • Integrated SEM imaging and chemical characterization
  • Enhanced automation to simplify workflows
  • Extended source lifetime and schedulable upgrades
Helios 6 HD FIB-SEM
  • High-volume TEM sample prep
  • Maximized system utilization
  • Failure analysis and metrology
Meridian EX System
  • Novel electron beam probing
  • 10x resolution versus optical
  • <5 nm node defect localization
Hydra Bio Plasma-FIB
  • Volume EM for cryo or resin-embedded samples with 4 switchable ion species
  • Millimeter-scale slicing with high z-precision with Spin Mill Bio Method
  • Integrated light microscope for correlative targeting
Iliad (S)TEM
  • Advanced integration of EELS and (S)TEM optics
  • Electrostatic beam blanker
  • High energy resolution electron source
Krios 5 Cryo-TEM
  • Fast, high-quality data collection
  • High workflow efficiency
  • Increased productivity and automation
  • Designed-in sustainability
Glacios 2 Cryo-TEM
  • High-resolution structure determination tool (<3Å)
  • Unparalleled ease-of-use through AI-enhanced automation
  • Flexible modes of operation standard: SPA, tomography, MicroED
Tundra Cryo-TEM
  • Structural information at biologically relevant resolution
  • Space efficient and cost effective
  • Easy, iterative sample optimization
  • Unique AI algorithms for streamlined data collection
Spectra Ultra TEM
  • New imaging and spectroscopy capabilities on the most beam sensitive materials
  • A leap forward in EDX detection with Ultra-X
  • Column designed to maintain sample integrity
Spectra 300 TEM
  • Highest-resolution structural and chemical information at the atomic level
  • Flexible high-tension range from 30-300 kV
  • Three lens condenser system
Spectra 200 TEM
  • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
  • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
  • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
Talos 12 TEM
  • Cell and tissue imaging
  • Tomography, CLEM, EDS
  • Negative stain imaging
  • Cryo-EM optimization
  • Accelerated nanoparticle characterization
Talos F200S TEM
  • Intuitive and easy-to-use automation software
  • Available with Super-X EDS for rapid quantitative chemical analysis
  • High-throughput with simultaneous multi-signal acquisition
Talos F200i TEM
  • Compact design with X-TWIN objective lens
  • Available with S-FEG, X-FEG, and X-CFEG
  • Flexible and fast EDS options for comprehensive elemental analysis
Talos F200X TEM
  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
  • X-FEG and X-CFEG available for the highest brightness and energy resolution
  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
Talos F200C TEM
  • High-contrast and high-quality TEM and STEM imaging
  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
  • Large pole piece gap and multiple in situ options
Talos L120C TEM
  • High versatility and stability
  • 4k x 4k Ceta CMOS camera for speed and large FOV
  • TEM magnification range of 25X to 650kX
  • EDS and STEM options for compositional analyses
Metrios AX TEM
  • Automation options to support quality, consistency, metrology, and reduced OPEX
  • Leverages machine learning for superior autofunctions and feature recognition
  • Workflows for both in-situ and ex-situ lamella preparation
Metrios 6 (S)TEM
  • Fully Automating Semiconductor Metrology Workflows
  • Recipe-Free Automation for (S)TEM Metrology
  • Efficient Chemical Analysis
Metrios 6S (S)TEM
  • Fully automated imaging/metrology
  • Automated TEM grid handling
  • Automated data connectivity
ExSolve WTP DualBeam
  • Can prepare site-specific, 20 nm thick lamellae on whole wafers up to 300 mm in diameter
  • Addresses needs requiring automated, high-throughput sampling at advanced technology nodes
Helios 5 EXL DualBeam
  • Low-voltage performance for high sample preparation quality
  • Ultra-high-resolution immersion-lens field-emission SEM column
  • Automated handling of 300 mm FOUP with EFEM (GEM300 compliant)
Helios 5 DualBeam
  • Fully automated, high-quality, ultra-thin TEM sample preparation
  • High throughput, high resolution subsurface and 3D characterization
  • Rapid nanoprototyping capabilities
Helios 5 PFIB DualBeam
  • Gallium-free STEM and TEM sample preparation
  • Multi-modal subsurface and 3D information
  • Next-generation 2.5 μA xenon plasma FIB column
Helios Hydra 5+ PFIB-SEM
  • 50% current increase for Xe and Ar beams, improving throughput for large volume cross-sectioning
  • Advanced automation for Ga-free TEM preparation
  • Ultra-low, 100eV SEM inspection for damage-free delayering
Helios Hydra 5 PFIB-SEM
  • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
  • Ga-free TEM sample preparation
  • Extreme high resolution SEM imaging
FIB-SEM and Laser Ablation
  • All three beams have same coincident point for accurate and repeatable cut placement
  • Millimeter-scale cross sections with up to 15,000x faster material removal than a typical FIB
  • Statistically relevant deep subsurface and 3D data analysis
Arctis Cryo-Plasma-FIB
  • Correlation to light microscopy and relocation in TEM
  • High-quality lamellae with consistent thickness
  • Automated high throughput and connectivity for cryo-tomography
Aquilos 2 Cryo-FIB
  • Automation enables production of multiple lamellas
  • Target and extract your structure of interest with lift-out nano-manipulator
  • 3D visualization for high-resolution tomography
Scios 3 FIB-SEM
  • Full support of magnetic and non-conductive samples
  • High throughput subsurface and 3D characterization
  • Advanced ease of use and automation capabilities
Axia ChemiSEM
  • Live quantitative elemental mapping
  • High fidelity scanning electron microscopy imaging
  • Flexible and easy to use, even for novice users
  • Easy maintenance
Verios 5 XHR SEM
  • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
  • Easy access to beam landing energies as low as 20 eV
  • Excellent stability with piezo stage as standard
Quattro ESEM
  • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
  • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
Prisma E SEM
  • Entry-level SEM with excellent image quality
  • Easy and quick sample loading and navigation for multiple samples
  • Compatible with a wide range of materials thanks to dedicated vacuum modes
Apreo 2 SEM
  • High-performance SEM for all-round nanometer or sub-nanometer resolution
  • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
  • Excellent performance at long working distance (10 mm)
VolumeScope 2 SEM
  • Isotropic 3D data from large volumes
  • High contrast and resolution in high and low vacuum modes
  • Simple switch between normal SEM use and serial block-face imaging
Phenom Pharos G2 Desktop FEG-SEM
  • FEG source with 1 - 20 kV acceleration voltage range
  • <2.0 nm (SE) and 3.0 nm (BSE) resolution @ 20 kV
  • Optional fully integrated EDS and SE detector
Phenom XL G2 Desktop SEM
  • For large samples (100x100 mm) and ideal for automation
  • <10 nm resolution and up to 200,000x magnification; 4.8 kV up to 20 kV acceleration voltage
  • Optional fully integrated EDS and BSE detector
Phenom ParticleX Battery Desktop SEM
  • Versatile solution for high-quality, in-house analysis
  • Automated system and analysis of multiple samples
  • Testing 10x faster
Phenom ProX G6 Desktop SEM
  • High performance desktop SEM with integrated EDS detector
  • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
  • Optional SE detector
Phenom Pro G6 Desktop SEM
  • High performance desktop SEM
  • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
  • Optional SE detector
Phenom Pure G6 Desktop SEM
  • Entry level desktop SEM
  • Resolution <15 nm; magnification up to 175,000x
  • Longlife CeB6 source
Phenom ParticleX GSR Desktop SEM
  • Dedicated automated GSR desktop SEM
  • Resolution <10 nm; magnification up to 200,000x
  • Longlife CeB6 source
Phenom ParticleX AM Desktop SEM
  • Versatile desktop SEM with automation software for Additive Manufacturing
  • Resolution <10 nm; magnification up to 200,000x
  • Optional SE detector
Phenom ParticleX TC Desktop SEM
  • Versatile desktop SEM with automation software for Technical Cleanliness
  • Resolution <10 nm; magnification up to 200,000x
  • Optional SE detector
Phenom ParticleX Steel Desktop SEM
  • SEM and EDS integrated
  • Ease of use
  • Sub-micrometer inclusions
ELITE System
  • Electrical defect localization
  • Thermal mapping
  • Advanced packaging analysis
Hyperion II System
  • Atomic Force Probing
  • Localize transistor faults
  • Integrated PicoCurrent (CAFM)
nProber IV System
  • Localize transistor and BEOL faults
  • Thermal nanoprobing (-40°C to 150°C)
  • Semi-automated operation
Meridian S System
  • Fault Diagnostic with Active Probe Technology
  • Static laser stimulation (SLS / OBIRCH) and photon emission options
  • Supports both micro-probing and probe card device stimulation
Meridian WS-DP System
  • High-sensitivity, low-noise, low-voltage photon emission detection with broadband DBX or InGaAs camera systems
  • Multi-wavelength laser scanning microscope for scan chain analysis, frequency mapping, transistor probing and isolation of faults
Meridian 7 System
  • Dynamic Optical Fault Isolation for 10nm node and below
  • High resolution visible and Infrared light
  • High-yield sample preparation to 5μm widely available
Meridian IV System
  • High sensitivity extended-wavelength DBX photon emission detection
  • Standard InGaAs photon emission detection
  • Laser Scanning Microscope with multiple wavelength options
Centrios Circuit Edit System
  • Superior image / milling resolution
  • Enhanced milling precision and control
  • Built on the Thermo Scientific Helios DualBeam platform
Centrios HX Circuit Edit System
  • Cutting-edge circuit editing for advanced semiconductor nodes with the latest FIB technology
  • Unparalleled milling precision and control with low landing energy to preserve sensitive circuits
  • Advanced navigation and ion beam placement accuracy
MK.4TE ESD and Latch-Up Test System
  • Rapid-relay-based operations—up to 2304 channels
  • Advanced device preconditioning with six separate vector drive levels
  • Fully compliant Latch-Up stimulus and device biasing
Orion3 Electrostatic Discharge Tester
  • Charged device model testing
  • Dual high resolution color cameras
  • Test densities to less than 0.4mm pitch
Celestron Test System
  • Wafer and package level TLP testing
  • High current TLP pulse generator
  • Can be interfaced with semiautomatic probers
  • Intuitive software for control and report generation
Pegasus ESD Test System
  • Testing per the latest industry standards
  • True system level ESD 150pF/330Ω network
  • 2 pin connection via wafer probes to any device
Nexsa
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
K-Alpha
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Vitrobot System
  • Fully Automated Sample vitrification
  • Blotting Device
  • Semi-Automated Grid Transfer
  • High Sample Throughput
CleanMill System
  • Broad ion beam milling and polishing
  • Wide acceleration voltage range
  • Cryogenic sample preparation
  • CleanConnect compatible
Amira Software
  • Support for multi-data/view/channel
  • Interactive high-quality visualization
  • Machine Learning-based segmentation
  • Intuitive recipe creation
Athena Software
  • Ensure traceability of images, data, metadata and experimental workflows
  • Simplify your imaging workflow
  • Improve collaboration
  • Secure and manage data access
Auto Slice & View 5 Software
  • Automated serial sectioning for DualBeam
  • Multi-modal data acquisition (SEM, EDS, EBSD)
  • On-the-fly editing capabilities
AutoScript 4 Software
  • improved reproducibility and accuracy
  • Unattended, high throughput imaging and patterning
  • Supported by Python 3.5-based scripting environment
AutoScript TEM Software
  • Provides a direct link between research needs and microscope automation
  • Enables improved reproducibility and accuracy
  • Focuses time on the microscope for higher throughput
AutoTEM 5 Software
  • Fully automated in situ S/TEM sample preparation
  • Support of top-down, planar and inverted geometry
  • Highly configurable workflow
  • Easy to use, intuitive user interface
Avizo Software
  • Support for multi-data/multi-view, multi-channel, time series, very large data
  • Advanced multi-mode 2D/3D automatic registration
  • Artifact reduction algorithms
CryoFlow Software
  • Secure data sharing for remote and collaborative projects
  • Efficient data management and connectivity for cryo-EM workflows
  • Interactive high-quality visualization for single particle analysis and cryo-electron tomography results
Smart EPU Software
  • Microscope-embedded solution for single particle data acquisition
  • Supports fully autonomous screening
  • Real time image analysis with AI-driven algorithms
Embedded CryoSPARC Live
  • Real-time, multidimensional feedback on sample quality and instrument settings
  • Adjust imaging and processing based on real-time information
  • Fully integrated with Smart EPU Software
iFast Software
  • Macro recorder for faster recipe creating
  • Runner for unattended overnight operation
  • Alignment tools: Image recognition and edge finding
Inspect 3D Software
  • Image processing tools and filters for cross-correlation
  • Feature tracking for image alignment
  • Algebraic reconstruction technique for iterative projection comparison
Maps Software
  • Acquire high-resolution images over large areas
  • Easily find regions of interest
  • Automate image acquisition process
NEXS Software
  • Auto-syncs position/magnification between NEXS and Circuit Edit system for a more seamless user experience
  • Connects to most Thermo Scientific tools used for EFA, PFA and Circuit Edit space
Pergeos Software
Rocks and Minerals
  • Visualize, process, and analyze rock imagery data
  • Perform multi-scale imaging analysis from pore to core
  • Calculate geological and petrophysical properties
Tomography 5 Software
  • Automated solution for 3D Cryo-EM tomography data acquisition
  • Automatic cassette mapping
  • Dose symmetric tilt scheme for optimal electron dose
Velox Software
  • Focus on getting your insights with all-in-one software
  • Interactive live graphical detector layout
  • Time resolved imaging and analytics – for best results in all experiments
3D Reconstruction
  • Intuitive user interface, maximum employability
  • Intuitive fully automated user interface
  • Based on 'shape from shading' technology, no stage tilt required
Elemental Mapping
  • Fast and reliable information on the distribution of elements within the sample or the selected line
  • Easily exported and reported results
FiberMetric
  • Save time by automated measurements
  • Fast and automated collection of all statistical data
  • View and measure micro and nano fibers with unmatched accuracy
Nanobuilder
  • CAD-based prototyping
  • Fully automated job execution, stage navigation, milling, and deposition
  • Automated alignment and drift control
ParticleMetric
  • Integrated software for online and offline analysis
  • Correlating particle features such as diameter, circularity, aspect ratio and convexity
  • Creating image datasets with Automated Image Mapping
Phenom Programming Interface
  • Customize your SEM to fit your workflow
  • Increase efficiency and save time with automated processes
  • Control imaging settings and stage navigation
PoroMetric
  • Correlate pore features such as area, aspect ratio, major and minor axis
  • Acquire images directly from the Desktop SEM
  • Statistical data with high-quality images
ProSuite
  • Automated collection of images
  • Real-time remote control
  • Standard applications included: Automated Image Mapping + Remote User Interface
Quartz PCI/CFR
  • SEM imaging traceability compliant with 21 CFR Part 11
  • Compatible with the Phenom XL and Phenom Pro desktop SEMs
  • Windows 10 64-bit operating system support
μHeater
  • Ultra-fast heating solution for in situ high resolution imaging
  • Fully integrated
  • Temperatures up to 1200 °C
μPolisher
  • Potential to enable large number of novel, unexplored applications
  • Very low energy milling
  • Small spot size for precise local surface treatment
Selectris and Selectris X Imaging Filters
  • Designed for high stability and atomic resolution imaging
  • Straightforward operation
  • Paired with the latest generation Thermo Scientific Falcon 4 Direct Electron Detector
Ceta D Camera
  • Optimum performance at any high tension (20–300 kV)
  • Compatible with post-column filters and spectrometers
  • Movie acquisition for dynamic studies
Falcon 4i Detector
  • High thorughput for more images per hour
  • Unsurpassed imaging quality with high DQE
  • Lossless data compression with EER
Charge Reduction Sample Holder
  • Up to 8 times higher magnification
  • Faster sample preparation
  • Non conductive samples can be imaged in their natural state
Electrical Feedthrough Sample Holder
  • Connect electrical probes to the sample for in situ measurements
  • Sample can be height adjusted from 0–25 mm manually
  • Measurements of probe currents
Eucentric Sample Holder
  • Eucentric tilting and compucentric rotation on a desktop SEM
  • Fast time-to-image with sample loading < 1 minute
  • Real-time 3D sample visualization module
Filter Inserts
  • Filter residue analysis and asbestos analysis
  • Available in two models which support 47 mm (1.85 inch) and 25 mm (1 inch) filters
  • Use on Phenom Desktop SEM
Metallurgical Sample Holder
  • Designed to support resin-mounted samples
  • Preferred solution for metallurgy and when working with inserts
  • Sample size up to 32 mm diameter and 30 mm height
Micro Tool Sample Holder
  • Quick and fast clamping
  • Tilting and rotation allow for easy sample positioning
  • No extra tooling required for sample loading
Motorized Tilt & Rotation Sample Holder
  • Tilt range -10° to +45°
  • Continuous 360° compucentric rotation
  • Controlled by dedicated Motion Control ProSuite application
Nebula Particle Disperser
  • Standard method for uniform dry powder dispersion
  • Avoids particle clusters
  • Used with Phenom Desktop SEM
Resin Mount Inserts
  • A unique sample holder concept
  • Available in 3 models for supporting standard sized samples of 25 mm (~1 inch), 32 mm (~1 ¼ inch) and 40 mm (~1 ½ inch) diameter
Standard Sample Holder
  • Compact stage allowing analysis of samples of up to 100 mm x 100 mm
  • Can be extended with 3 types of resin or metallurgical mount inserts
  • Used with Phenom Desktop SEM
Sample Holder Inserts
  • Faster cross-sectional imaging of coatings and multilayer samples
  • Easy clamping without the need for screws or extra tools
  • Eliminates the need for screws and tools to clamp the sample
 Download datasheet
Tensile Sample Holder
  • Determine batch quality
  • Determine manufacturing consistencys
  • Aid the design process
Temperature Controlled Sample Holder
  • Temperature range -25 °C to +50 °C
  • Temperature accuracy ±1.5 °C
  • Temperature display resolution 0.1 °C
Resources
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards